Specifications
- Color:Natural
- Description:8-Strip flat caps for PCR tubes and plates, DNase, RNase and DNA free
- Cat. no.:104028-678
- Packaging:125 strips of 81000 per pack
Specifications
About this item
Ensure reliable sealing with Alkali Scientific™ ProCycle™ 8-Strip flat caps for PCR tubes and plates, featuring Clear material and DNase, RNase and DNA Free construction, available in packs of 125 strips of 8 and cases of 1000.
- Exceptional thermal conductivity for precise thermal cycling
- Optically clear
Maintain sample integrity and prevent contamination with ProCycle™ 8-Strip flat caps for PCR tubes and plates. These clear caps are designed to securely seal PCR tubes and plates, providing a tight and leak-proof seal to prevent sample evaporation and contamination. With DNase, RNase and DNA-free certification, these caps ensure the integrity of nucleic acid samples throughout the PCR process. Each pack contains 125 strips of 8 caps, providing ample supply for your laboratory needs. Designed for convenience and reliability, ProCycle™ 8-Strip flat caps are essential accessories for your molecular biology experiments.