ContactUsLinkComponent
ProCycle™ Flat Caps for 8-PCR Strip Tubes
  104027-262
undefined
ProCycle™ Flat Caps for 8-PCR Strip Tubes
  104027-262
 :  PCRF-8-CAP
  • Color:
    Clear
  • Description:
    8-Strip flat caps, for RT PCR, use with PCR tubes or PCR plates
  • Cat. no.:
    104027-262
  • Packaging:
    125 strips of 8 of 8
Secure PCR strip tubes with Alkali Scientific™ ProCycle™ flat caps, available in packs of 125 for convenient sealing.
  • Exceptional thermal conductivity for precise thermal cycling
  • Optically clear
Ensure reliable sealing and protection of PCR strip tubes with ProCycle™ flat caps. These caps provide a secure and leak-proof seal, preventing sample evaporation and contamination during PCR amplification. Featuring a low-profile design for convenient storage and handling, the flat caps offer reliable sample protection while maintaining ease of use. Designed to fit securely onto PCR strip tubes, these caps ensure sample integrity and reproducibility in your molecular biology experiments.