ContactUsLinkComponent
ProCycle™ 0.2 ml Clear 8-Strip Tubes with Attached Hinged Domed Cap
ProCycle™ 0.2 ml Clear 8-Strip Tubes with Attached Hinged Domed Cap
Catalog # 104027-048
Supplier:  Alkali Scientific
undefined
ProCycle™ 0.2 ml Clear 8-Strip Tubes with Attached Hinged Domed Cap
Catalog # 104027-048
Supplier:  Alkali Scientific
Supplier Number:  PC7066
Specifications
  • Color:
    Clear
  • Description:
    8-Strip PCR tubes with attached hinged domed cap strip, 0.2 ml
  • Cat. no.:
    104027-048
  • Packaging:
    125×8, 1000 per pack
Specifications
About this item
Enhance sample visibility with Alkali Scientific™ ProCycle™ 0.2 ml clear 8-Strip tubes featuring attached hinged domed caps, available in packs of 1000.
  • 0.2 ml 8-Strip PCR tubes with attached dome caps
  • Caps are optically clear for qPCR
  • Ultra-thin walls for uniformity and consistency
  • Positive sealing eliminates any evaporation
  • Fits standard spacing on 8-channel pipettes
  • Certified RNAse, DNAse and pyrogen free
Streamline laboratory operations with the ProCycle™ 0.2 ml clear 8-Strip PCR tubes and attached hinged domed caps. Designed for rapid thermal transfer and secure sealing, each pack of 1000 tubes ensures uninterrupted experimentation, allowing researchers to accelerate their scientific progress with ease and efficiency. With their transparent design, researchers can easily monitor samples while enjoying the convenience of attached hinged domed caps.