ContactUsLinkComponent
ProCycle™ 0.2 ml 8-Strip PCR Tubes with Separate Dome Strip Caps
ProCycle™ 0.2 ml 8-Strip PCR Tubes with Separate Dome Strip Caps
Catalog # 104027-704
Supplier:  Alkali Scientific
undefined
ProCycle™ 0.2 ml 8-Strip PCR Tubes with Separate Dome Strip Caps
Catalog # 104027-704
Supplier:  Alkali Scientific
Supplier Number:  PC7063
Specifications
  • Description:
    8-Strip PCR tubes with separate dome strip caps, 0.2 ml
  • Cat. no.:
    104027-704
  • Packaging:
    125x8, 1000 per pack
Specifications
About this item
Efficiently conduct PCR experiments with Alkali Scientific™ ProCycle™ 0.2 ml 8-Strip PCR tubes featuring separate dome strip caps, available in packs of 1000.
  • 0.2 ml strip tubes with separate domed caps
  • Ultra-thin walls
  • Positive sealing
  • Fits standard spacing for 8-channel pipettes
  • Certified RNAse, DNAse and pyrogen free
Experience versatility and reliability at its finest with the ProCycle™ 0.2 ml 8-Strip PCR tubes and separate dome strip caps. Engineered for rapid thermal transfer and optimal sample integrity, these tubes and caps guarantee consistent performance, empowering researchers to achieve reproducible results with ease, even in high-throughput PCR assays. With a generous pack size of 1000 tubes, researchers can strea mline their experiments without compromising on reliability or efficiency.