About this item
Ensure precise electroporation with individually wrapped Alkali Scientific™ Electroporation Cuvettes, available in a pack of 50, for consistent results.
- Manufactured from high quality durable polycarbonate material which has been tested to optimize transfection efficiencies
- Precision built to give exacting tolerances ensuring reproducible results
- Polished aluminum plates reduce static and arcing for optimum performance
- Individually wrapped and gamma irradiated providing guaranteed sterility
- Compatible with most Electroporation devices
- Individually color-coded cuvette caps identify gap widths of 1 mm, 2 mm, and 4 mm
CellPro™ Electroporation Cuvettes are meticulously designed and manufactured to meet the demanding needs of genetic transformation experiments. Each cuvette is individually wrapped to maintain sterility and integrity until ready for use, ensuring reliability and consistency in experimental results. With their high-quality construction and precise engineering, these cuvettes provide researchers with a robust and dependable tool for conducting electroporation procedures. Whether used for gene editing, transfection, or other molecular biology applications, CellPro™ Electroporation Cuvettes offer researchers peace of mind and confidence in achieving successful outcomes in their genetic engineering endeavors.