ContactUsLinkComponent
LabKlenz™ 120 Alkaline Detergents
LabKlenz™ 120 Alkaline Detergents
Catalog # CA89089-206
Supplier:  Steris
undefined
LabKlenz™ 120 Alkaline Detergents
Catalog # CA89089-206
Supplier:  Steris
Supplier Number:  1L1205
Specifications
  • Size:
    19 L (5 gal.)
  • Cat. no.:
    CA89089-206
  • Packaging:
    Plastic pail
  • Supplier No.:
    1L1205
Specifications
About this item
LabKlenz™ 120 Detergent is a phosphate-free, liquid, alkaline detergent for use in automated laboratory glassware washing applications. It is compatible with a variety of substrates including stainless steel, mild steel, and copper. It is extremely free rinsing to meet critical laboratory requirements.
  • Efficacy - Effective removal of laboratory residues in both soft and hard water
  • Flexibility - Mild alkalinity for better materials compatibility
  • Cost savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
  • Environmentally friendly - phosphate-free formulation with biodegradable surfactants
  • Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
LabKlenz™ 120 Detergent is specially formulated to meet the unique cleaning and materials compatibility requirements found in laboratory glassware washing applications.