Order Entry
Canada
ContactUsLinkComponent
Human Recombinant UCHL5 (His tag), Active (from E. coli)
Human Recombinant UCHL5 (His tag), Active (from E. coli)
Supplier:  Sino Biological
New Product
Certificates
Human Recombinant UCHL5 (His tag), Active (from E. coli)
Supplier:  Sino Biological
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
  • Protein synonyms:
    CGI-70|UCH-L5|INO80R|UCH37
  • Protein/peptide name:
    UCHL5
  • Molecular weight:
    38 kDa
  • Formulation:
    Supplied as sterile 50 mM sodium phosphate, pH 7.5, 300 mM NaCl, 50 - 200 mM imidazole, 0 - 0.1 mM PMSF, 0.2 - 1.0 mM DTT, 10 - 25% glycerol
  • Tested applications:
    WB, Activity Assay

Specifications

Related Information

Frequently Bought Together