Human Recombinant SAE1/UBA2 Complex (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
New Product
Certificates
About this item
Recombinant full length human SAE1 (UBLE1A) and human UBA2 (SAE2) were co-expressed by baculovirus in Sf9 insect cells using an N-terminal His tag on SAE1 and GST tag on UBA2.
- Shipping temperature: Dry ice
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Human
- Storage conditions:Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
- Protein/peptide name:SAE1 & UBA2
- Molecular weight:~39 kDa (SAE1); ~115 kDa (UBA2)
- Formulation:Supplied as sterile 50 mM Tris-HCl, pH 7.5, 50 - 300 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 - 1.0 mM DTT, 0 - 0.1 mM PMSF, 10 - 25% glycerol
- Tested applications:WB, Ubiquitin Enzyme Assay
Specifications
Related Information
Frequently Bought Together