Order Entry
Canada
ContactUsLinkComponent
 
Human Recombinant Tau-441 (A152T) (from E. coli)
New Product
 
Human Recombinant Tau-441 (A152T) (from E. coli)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
  • Protein synonyms:
    PPP1R103|FTDP-17|MAPT|TAU|PPND|MAPTL|MTBT1|MTBT2|MSTD|DDPAC
  • Protein/peptide name:
    MAPT
  • Molecular weight:
    64 kDa (45.8 kDa by MW calculation)
  • Formulation:
    Supplied as sterile 50 mM Tris-HCl pH 7.5, 50 - 300 mM NaCl, 1 mM DTT, 10 - 25% glycerol

 

Frequently Bought Together