Human Recombinant Tau-441 (151-441) (from E. coli)
Supplier: Sino Biological
New Product
Certificates
About this item
Recombinant human tag-free Tau-441 (151-441) was expressed in E. coli cells.
- Shipping temperature: Dry ice
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
- Protein synonyms:PPP1R103|FTDP-17|MAPT|TAU|PPND|MAPTL|MTBT1|MTBT2|MSTD|DDPAC
- Protein/peptide name:MAPT
- Molecular weight:~34 kDa
- Formulation:Supplied as sterile 50 mM Tris-HCl pH 7.5, 50 - 300 mM NaCl, 1 mM DTT, 10 - 25% glycerol
Specifications
Frequently Bought Together