Human Recombinant AKT2 (His tag), Unactive (from Baculovirus-Insect Cells)
Supplier: Sino Biological
Certificates
About this item
Recombinant human AKT2 (142-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
- Shipping temperature: Dry ice
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Human
- Storage conditions:Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
- Protein synonyms:RAC-BETA|PKBB|PRKBB|PKBBETA|HIHGHH
- Protein/peptide name:AKT2
- Molecular weight:~41 kDa
- Formulation:Supplied as sterile 50 mM sodium phosphate, pH 7.5, 300 mM NaCl, 50 - 200 mM imidazole, 0 - 0.1 mM PMSF, 0.2 - 1.0 mM DTT, 10 - 25% glycerol
Specifications
Related Information
Frequently Bought Together