Human Recombinant ATF1 (GST tag) (from Baculovirus-Insect Cells)
New Product
Recombinant full length human ATF1 was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag.
- Shipping temperature: Dry ice
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Human
- Storage conditions:Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
- Protein synonyms:FUS/ATF-1|TREB36|EWS-ATF1
- Protein/peptide name:ATF1
- Molecular weight:~63 kDa
- Formulation:Supplied as sterile 50 mM Tris-HCl, pH 7.5, 50 - 300 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 - 1.0 mM DTT, 0 - 0.1 mM PMSF, 10 - 25% glycerol
Frequently Bought Together