Orders LinkContactUsLinkComponent
Automatic Liquid Sampler Kit
Automatic Liquid Sampler Kit
Basic automatic liquid sampler supply kit
Catalog # AG07673-60840
Supplier #:  07673-60840
Automatic Liquid Sampler Kit
Catalog # AG07673-60840
Automatic Liquid Sampler Kit
Basic automatic liquid sampler supply kit
Price
$2,490.00
Log in for availability
Kit of 1
Product Details & Documents
Basic supplies kit (7673A), includes six 10 μl 23/42/cone syringes, 4 ml vial with diffusion cap (144/pack), 2 ml screw-top vials with caps (1000/pack), inlet septa (100/pack), vial rack (5/pack).
Specifications
  • Description:
    Basic automatic liquid sampler supply kit