Erlenmeyer Flasks, PETG, Flat Bottom
Supplier: NEST SCIENTIFIC
Jump to:
About this item
Ideal for shaker culture applications. PETG Erlenmeyer flasks are manufactured in 100000 grade cleaning room with USP 6 grade raw materials.
- PETG Bottle and HDPE cap
- Sterilized by E-Beam
- Nonpyrogenic
- Packaged in sterile bags
Vent caps with 0.22 µm hydrophobic filters for gas exchange without contamination.
Certifications: ISO 9001 and 13485 certified.
8 Options Available Below
Product Details & Documents
Ideal for shaker culture applications. PETG Erlenmeyer flasks are manufactured in 100000 grade cleaning room with USP 6 grade raw materials.
- PETG Bottle and HDPE cap
- Sterilized by E-Beam
- Nonpyrogenic
- Packaged in sterile bags
Vent caps with 0.22 µm hydrophobic filters for gas exchange without contamination.
Certifications: ISO 9001 and 13485 certified.
Product Family Options
With plug seal capped
Product Information
- CapacitySterilityHeightNeck O.D.AvailabilityPrice
- 77508-804Conical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 66 mm, 24 per caseConical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 66 mm, 24 per case125 mlSterile106.8 mm34 mmCase of 24$197.71Each$197.71Supplier Number: 781001
- 77508-820Conical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 83 mm, 12 per caseConical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 83 mm, 12 per case250 mlSterile137.8 mm34 mmCase of 12$138.20Each$138.20Supplier Number: 782001
- 77508-836Conical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 101 mm, 12 per caseConical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 101 mm, 12 per case500 mlSterile175.8 mm39 mmCase of 12$186.28Each$186.28Supplier Number: 783001
- 77508-852Conical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 127 mm, 6 per caseConical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 127 mm, 6 per case1000 mlSterile213.3 mm39 mmCase of 6$153.54Each$153.54Supplier Number: 784001
With vented cap
Product Information
- CapacitySterilityHeightNeck O.D.AvailabilityPrice
- 77508-806Conical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 66 mm, 24 per caseConical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 66 mm, 24 per case125 mlSterile106.8 mm34 mmCase of 24$209.04Each$209.04Supplier Number: 781011
- 77508-822Conical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 83 mm, 12 per caseConical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 83 mm, 12 per case250 mlSterile137.8 mm34 mmCase of 12$142.44Each$142.44Supplier Number: 782011
- 77508-838Conical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 101 mm, 12 per caseConical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 101 mm, 12 per case500 mlSterile175.8 mm39 mmCase of 12$195.77Each$195.77Supplier Number: 783011
- 77508-854Conical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 127 mm, 6 per caseConical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 127 mm, 6 per case1000 mlSterile213.3 mm39 mmCase of 6$162.76Each$162.76Supplier Number: 784011