Orders LinkContactUsLinkComponent
High Efficiency Erlenmeyer Flasks, Polycarbonate, Flat Bottom
High Efficiency Erlenmeyer Flasks, Polycarbonate, Flat Bottom
Supplier:   NEST SCIENTIFIC
High Efficiency Erlenmeyer Flasks, Polycarbonate, Flat Bottom
Supplier:  NEST SCIENTIFIC
Certificates
High Efficiency Erlenmeyer Flasks, Polycarbonate, Flat Bottom
8 Options Available Below
Product Details & Documents
High-efficiency, large-volume culture flasks allow cells to show strong viability with large expression amount of proteins in the culture of mammalian cells and insect cells.
  • PC bottle, which meets the USP Class 6 Standard, with high transparency, has strong impact resistance, oxidation resistance and can withstand a high temperature up to 121 ℃
  • There’s a scale made by injection molding on the flask body to facilitate observing the filled liquid volume
  • Sterilized by E-beam, SAL= 10⁻⁶
  • Non-pyrogenic, DNase/Rnase free
  • Individually packaged in sterile bag
During the culturing process, the use rate of the shaker is significantly increased, and the survival rate and
viability of cells are both dramatically elevated.
Product Family Options
With sealed cap
Product Information
  • Capacity
    Sterility
    Availability
    Price
  • 77508-868
    PC High efficient Erlenmeyer flask, flat bottom
    PC High efficient Erlenmeyer flask, flat bottom
    2 L
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $268.40
    $268.40
    Log in for availability
    Each
    Each - $268.40
    $268.40
    Case of 4
       $268.40
    Each
       $268.40
    Supplier Number: 785101
  • 77508-884
    PC High efficient Erlenmeyer flask, flat bottom
    PC High efficient Erlenmeyer flask, flat bottom
    3 L
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $458.74
    $458.74
    Log in for availability
    Each
    Each - $458.74
    $458.74
    Case of 4
       $458.74
    Each
       $458.74
    Supplier Number: 786101
  • 77508-892
    PC High efficient Erlenmeyer flask, flat bottom, wide mouth
    PC High efficient Erlenmeyer flask, flat bottom, wide mouth
    3 L
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $458.74
    $458.74
    Log in for availability
    Each
    Each - $458.74
    $458.74
    Case of 4
       $458.74
    Each
       $458.74
    Supplier Number: 786501
  • 77508-904
    PC High efficient Erlenmeyer flask, flat bottom
    PC High efficient Erlenmeyer flask, flat bottom
    5 L
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $680.66
    $680.66
    Log in for availability
    Each
    Each - $680.66
    $680.66
    Case of 4
       $680.66
    Each
       $680.66
    Supplier Number: 787001
With vented cap
Product Information
  • Capacity
    Sterility
    Availability
    Price
  • 77508-872
    PC High efficient Erlenmeyer flask, flat bottom
    PC High efficient Erlenmeyer flask, flat bottom
    2 L
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $277.15
    $277.15
    Log in for availability
    Each
    Each - $277.15
    $277.15
    Case of 4
       $277.15
    Each
       $277.15
    Supplier Number: 785111
  • 77508-888
    PC High efficient Erlenmeyer flask, flat bottom
    PC High efficient Erlenmeyer flask, flat bottom
    3 L
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $464.39
    $464.39
    Log in for availability
    Each
    Each - $464.39
    $464.39
    Case of 4
       $464.39
    Each
       $464.39
    Supplier Number: 786111
  • 77508-896
    PC High efficient Erlenmeyer flask, flat bottom, wide mouth
    PC High efficient Erlenmeyer flask, flat bottom, wide mouth
    3 L
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $468.72
    $468.72
    Log in for availability
    Each
    Each - $468.72
    $468.72
    Case of 4
       $468.72
    Each
       $468.72
    Supplier Number: 786511
  • 77508-908
    PC High efficient Erlenmeyer flask, flat bottom
    PC High efficient Erlenmeyer flask, flat bottom
    5 L
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $694.48
    $694.48
    Log in for availability
    Each
    Each - $694.48
    $694.48
    Case of 4
       $694.48
    Each
       $694.48
    Supplier Number: 787011