Orders LinkContactUsLinkComponent
Transfer Caps for High Efficient Erlenmeyer Flasks, Multifunction Liquid System | Avantor
Transfer Caps for High Efficient Erlenmeyer Flasks, Multifunction Liquid System
Supplier:   NEST SCIENTIFIC
Transfer Caps for High Efficient Erlenmeyer Flasks, Multifunction Liquid System
Supplier:  NEST SCIENTIFIC
Certificates
Transfer Caps for High Efficient Erlenmeyer Flasks, Multifunction Liquid System
14 Options Available Below
Product Details & Documents
Transfer caps for high efficient erlenmeyer flask.
  • Compatible with 2/3/5 L high efficient Erlenmeyer flasks
  • The liquid inlet tubing is provided with a PTFE needle filter, which solves the issue of liquid remaining in the tubing during the
  • feeding process
Different from bi-directional liquid transfer cap, the multifunctional transfer cap can be directly placed in an incubator for culture after the liquid transfer is completed. It can reach a large air flux. The quick sampling connector is composed of a sampling nozzle and a one-way valve, which can prevent the liquid from flowing backwards during the sampling process and ensure the aseptic sampling.
Product Family Options
Product Information
  • For use with
    Length
    Ø
    Availability
    Price
  • 77508-994
    Multifunction liquid transfer cap with 20 cm² filter membrane area,heat seal connecter
    Multifunction liquid transfer cap with 20 cm² filter membrane area,heat seal connecter
    2 L Flasks
    50 cm (External)
    24 cm (Internal)
    External liquid transfer tube: 3.2 mm (int.Ø); 6.4 mm (ext.Ø)
    Internal liquid transfer tube: 4 mm (int.Ø); 6 mm (ext.Ø)
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $1,542.55
    $1,542.55
    Log in for availability
    Each
    Each - $1,542.55
    $1,542.55
    Case of 4
       $1,542.55
    Each
       $1,542.55
    Supplier Number: C10111-AJE052B
  • 77508-996
    Multifunction liquid transfer cap with 4.5 cm² filter membrane area,heat seal connecter
    Multifunction liquid transfer cap with 4.5 cm² filter membrane area,heat seal connecter
    2 L Flasks
    50 cm (External)
    24 cm (Internal)
    External liquid transfer tube: 6.4 mm (int.Ø); 11.1 mm (ext.Ø)
    Internal liquid transfer tube: 4 mm (int.Ø); 6 mm (ext.Ø)
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $1,542.55
    $1,542.55
    Log in for availability
    Each
    Each - $1,542.55
    $1,542.55
    Case of 4
       $1,542.55
    Each
       $1,542.55
    Supplier Number: C10111-DZB050B
  • 77508-998
    Multifunction liquid transfer cap with 4.5 cm² filter membrane area,MPC male connecter
    Multifunction liquid transfer cap with 4.5 cm² filter membrane area,MPC male connecter
    2 L Flasks
    100 cm (External)
    24 cm (Internal)
    External liquid transfer tube: 4.8 mm (int.Ø); 9.53 mm (ext.Ø)
    Internal liquid transfer tube: 4 mm (int.Ø); 6 mm (ext.Ø)
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $1,542.55
    $1,542.55
    Log in for availability
    Each
    Each - $1,542.55
    $1,542.55
    Case of 4
       $1,542.55
    Each
       $1,542.55
    Supplier Number: C10111-GBB100B
  • 77509-016
    Multifunction liquid transfer cap with 20 cm² filter membrane area,heat seal connecter
    Multifunction liquid transfer cap with 20 cm² filter membrane area,heat seal connecter
    3 L Flasks
    50 cm (External)
    28 cm (Internal)
    External liquid transfer tube: 3.2 mm (int.Ø); 6.4 mm (ext.Ø)
    Internal liquid transfer tube: 4 mm (int.Ø); 6 mm (ext.Ø)
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $1,635.54
    $1,635.54
    Log in for availability
    Each
    Each - $1,635.54
    $1,635.54
    Case of 4
       $1,635.54
    Each
       $1,635.54
    Supplier Number: C10211-AJE052B
  • 77509-018
    Multifunction liquid transfer cap with 4.5 cm² filter membrane area,heat seal connecter
    Multifunction liquid transfer cap with 4.5 cm² filter membrane area,heat seal connecter
    3 L Flasks
    50 cm (External)
    28 cm (Internal)
    External liquid transfer tube: 6.4 mm (int.Ø); 11.1 mm (ext.Ø)
    Internal liquid transfer tube: 4 mm (int.Ø); 6 mm (ext.Ø)
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $940.76
    $940.76
    Log in for availability
    Each
    Each - $940.76
    $940.76
    Case of 4
       $940.76
    Each
       $940.76
    Supplier Number: C10211-DZB050B
  • 77509-020
    Multifunction liquid transfer cap with 4.5 cm² filter membrane area,MPC male connecter
    Multifunction liquid transfer cap with 4.5 cm² filter membrane area,MPC male connecter
    3 L Flasks
    100 cm (External)
    28 cm (Internal)
    External liquid transfer tube: 4.8 mm (int.Ø); 9.53 mm (ext.Ø)
    Internal liquid transfer tube: 4 mm (int.Ø); 6 mm (ext.Ø)
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $1,635.54
    $1,635.54
    Log in for availability
    Each
    Each - $1,635.54
    $1,635.54
    Case of 4
       $1,635.54
    Each
       $1,635.54
    Supplier Number: C10211-GBB100B
  • 77509-040
    Multifunction liquid transfer cap with 20 cm² filter membrane area,heat seal connecter
    Multifunction liquid transfer cap with 20 cm² filter membrane area,heat seal connecter
    3 L Flasks, wide mouth
    50 cm (External)
    28 cm (Internal)
    External liquid transfer tube: 3.2 mm (int.Ø); 6.4 mm (ext.Ø)
    Internal liquid transfer tube: 4 mm (int.Ø); 6 mm (ext.Ø)
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $1,622.42
    $1,622.42
    Log in for availability
    Each
    Each - $1,622.42
    $1,622.42
    Case of 4
       $1,622.42
    Each
       $1,622.42
    Supplier Number: C10311-AJE052B
  • 77509-042
    Multifunction liquid transfer cap with 4.5 cm² filter membrane area,heat seal connecter
    Multifunction liquid transfer cap with 4.5 cm² filter membrane area,heat seal connecter
    3 L Flasks, wide mouth
    50 cm (External)
    28 cm (Internal)
    External liquid transfer tube: 6.4 mm (int.Ø); 11.1 mm (ext.Ø)
    Internal liquid transfer tube: 4 mm (int.Ø); 6 mm (ext.Ø)
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $940.76
    $940.76
    Log in for availability
    Each
    Each - $940.76
    $940.76
    Case of 4
       $940.76
    Each
       $940.76
    Supplier Number: C10311-DZB050B
  • 77509-044
    Multifunction liquid transfer cap with 4.5 cm² filter membrane area,MPC male connecter
    Multifunction liquid transfer cap with 4.5 cm² filter membrane area,MPC male connecter
    3 L Flasks, wide mouth
    100 cm (External)
    28 cm (Internal)
    External liquid transfer tube: 4.8 mm (int.Ø); 9.53 mm (ext.Ø)
    Internal liquid transfer tube: 4 mm (int.Ø); 6 mm (ext.Ø)
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $1,622.42
    $1,622.42
    Log in for availability
    Each
    Each - $1,622.42
    $1,622.42
    Case of 4
       $1,622.42
    Each
       $1,622.42
    Supplier Number: C10311-GBB100B
  • 77509-062
    Multifunction liquid transfer cap with 20 cm² filter membrane area,heat seal connecter
    Multifunction liquid transfer cap with 20 cm² filter membrane area,heat seal connecter
    5 L Flasks
    90 cm (External)
    34 cm (Internal)
    External liquid transfer tube: 3.2 mm (int.Ø); 6.4 mm (ext.Ø)
    Internal liquid transfer tube: 4 mm (int.Ø); 6 mm (ext.Ø)
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $1,622.42
    $1,622.42
    Log in for availability
    Each
    Each - $1,622.42
    $1,622.42
    Case of 4
       $1,622.42
    Each
       $1,622.42
    Supplier Number: C10411-AJE092B
  • 77509-064
    Multifunction liquid transfer cap with 4.5 cm² filter membrane area,heat seal connecter
    Multifunction liquid transfer cap with 4.5 cm² filter membrane area,heat seal connecter
    5 L Flasks
    90 cm (External)
    34 cm (Internal)
    External liquid transfer tube: 6.4 mm (int.Ø); 11.1 mm (ext.Ø)
    Internal liquid transfer tube: 4 mm (int.Ø); 6 mm (ext.Ø)
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $1,112.88
    $1,112.88
    Log in for availability
    Each
    Each - $1,112.88
    $1,112.88
    Case of 4
       $1,112.88
    Each
       $1,112.88
    Supplier Number: C10411-DZB090B
  • 77509-066
    Multifunction liquid transfer cap with 4.5 cm² filter membrane area,MPC male connecter
    Multifunction liquid transfer cap with 4.5 cm² filter membrane area,MPC male connecter
    5 L Flasks
    100 cm (External)
    34 cm (Internal)
    External liquid transfer tube: 4.8 mm (int.Ø); 9.53 mm (ext.Ø)
    Internal liquid transfer tube: 4 mm (int.Ø); 6 mm (ext.Ø)
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $1,622.42
    $1,622.42
    Log in for availability
    Each
    Each - $1,622.42
    $1,622.42
    Case of 4
       $1,622.42
    Each
       $1,622.42
    Supplier Number: C10411-GBB100B
  • 77509-058
    Inverted lquid transfer cap
    Inverted lquid transfer cap
    3 L Flasks, wide mouth
    50 cm
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $768.51
    $768.51
    Log in for availability
    Each
    Each - $768.51
    $768.51
    Case of 4
       $768.51
    Each
       $768.51
    Supplier Number: C10331-AZC050B
  • 77509-122
    Sterile PETG square storage bottle bi-directional transfer cap with ¹/₄" TPE tube, female Luer connector with male sealing plug vent filter
    Sterile PETG square storage bottle bi-directional transfer cap with ¹/₄" TPE tube, female Luer connector with male sealing plug vent filter
    2 L Flasks
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $1,174.84
    $1,174.84
    Log in for availability
    Each
    Each - $1,174.84
    $1,174.84
    Case of 4
       $1,174.84
    Each
       $1,174.84
    Supplier Number: C501AA-BHC060B
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.