Order Entry
Canada
ContactUsLinkComponent
KIMAX® Heavy-Duty Centrifuge Tube, Graduated, with Flathead PTFE [ST] Stopper, Kimble Chase
KIMAX® Heavy-Duty Centrifuge Tube, Graduated, with Flathead PTFE [ST] Stopper, Kimble Chase
Catalog # 21054-190
undefined
KIMAX® Heavy-Duty Centrifuge Tube, Graduated, with Flathead PTFE [ST] Stopper, Kimble Chase
Catalog # 21054-190
Supplier Number:  45176-50

Specifications

  • Volume:
    50 ml
  • Cat. no.:
    21054-190
  • O.D.×L:
    29×155 mm
  • Supplier No.:
    45176-50

Specifications

About this item

Tube is calibrated to contain and includes a flathead, PTFE [ST]16 stopper.

  • Marking spot is blasted on both sides
  • Scale and legend are printed in permanent brown stain

Tube is graduated from 0 to 10 ml in 0.5 ml increments and from 10 to 50 ml in 1 ml increments.

Caution: Do not centrifuge with stopper in tube.