ContactUsLinkComponent
Pierce™ Protein Conjugate, Fluorescein, Biotinylated, Thermo Scientific
  CAPI22030
 :  
Pierce™ Protein Conjugate, Fluorescein, Biotinylated, Thermo Scientific
  CAPI22030
 :  22030
 :  
  • Description:
    Fluorescein, Biotin-Conjugated, Binds to Streptavidin; Produces a Greenish-Yellow Signal
  • Size:
    5 mg
  • Storage temperature:
    Store at 4 °C
  • Ex/Em:
    494/518 nm
  • Cat. no.:
    CAPI22030
  • Supplier No.:
    22030
Biotin-Fluorescein is a heterobifunctional reagent containing a biotin moiety and a fluorescein group separated by a long-chain hydrophilic polyethylene glycol (PEG) spacer arm.
Biotin-Fluorescein is used to label biotin binding protein molecules including avidin and streptavidin, which each have four biotin-binding sites. The resulting fluorescent biotin-binding protein can then amplify a signal generated in assays that use avidin-biotin detection systems.