Order Entry
Canada
Orders LinkContactUsLinkComponent
Inverted Liquid Transfer Cap System

Inverted Liquid Transfer Cap System

Supplier:   NEST SCIENTIFIC
Inverted Liquid Transfer Cap System
Supplier:  NEST SCIENTIFIC
Certificates
Inverted Liquid Transfer Cap System

7 Options Available Below

Product Details & Documents

Transfer caps for high efficient erlenmeyer flask.

  • Closed system reduces the risks of contamination during liquid transfer
  • One-piece construction of caps and connectors reduces the risk of leakage and media residue
  • High-quality materials and smooth inner wall of the tubing provide an excellent transfer performance
  • Sterilized by E-beam, SAL=10-6
  • Endotoxin-free, and no ingredients of animal origin

Caps of various diameters are available and aseptic welding of liquid inlet tubing under normal conditions is supported.

Packaging: Individually packaged in sterile bag

Product Family Options

Product Information

  • For use with
    Length
    Ø
    Sterility
    Availability
    Price
  • 2 L Flasks
    50 cm (External)
    24 cm (Internal)
    External liquid transfer tube: 3.2 mm (int.Ø); 6.4 mm (ext.Ø)
    Internal liquid transfer tube: 2 mm (int.Ø); 3 mm (ext.Ø)
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $768.51
    $768.51
    Log in for availability
    Each
    Each - $768.51
    $768.51
    Case of 4
       $768.51

    Each
       $768.51

    Supplier Number: C10131-AZC050B
  • 2 L Flasks
    50 cm (External)
    24 cm (Internal)
    External liquid transfer tube: 6.4 mm (int.Ø); 11.1 mm (ext.Ø)
    Internal liquid transfer tube: 2 mm (int.Ø); 3 mm (ext.Ø)
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $940.76
    $940.76
    Log in for availability
    Each
    Each - $940.76
    $940.76
    Case of 4
       $940.76

    Each
       $940.76

    Supplier Number: C10133-DZC050B
  • 3 L Flasks
    50 cm (External)
    28 cm (Internal)
    External liquid transfer tube: 3.2 mm (int.Ø); 6.4 mm (ext.Ø)
    Internal liquid transfer tube: 2 mm (int.Ø); 3 mm (ext.Ø)
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $768.51
    $768.51
    Log in for availability
    Each
    Each - $768.51
    $768.51
    Case of 4
       $768.51

    Each
       $768.51

    Supplier Number: C10231-AZC050B
  • 3 L Flasks
    50 cm (External)
    28 cm (Internal)
    External liquid transfer tube: 6.4 mm (int.Ø); 11.1 mm (ext.Ø)
    Internal liquid transfer tube: 2 mm (int.Ø); 3 mm (ext.Ø)
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $940.76
    $940.76
    Log in for availability
    Each
    Each - $940.76
    $940.76
    Case of 4
       $940.76

    Each
       $940.76

    Supplier Number: C10233-DZC050B
  • 3 L Flasks, wide mouth
    50 cm (External)
    28 cm (Internal)
    External liquid transfer tube: 6.4 mm (int.Ø); 11.1 mm (ext.Ø)
    Internal liquid transfer tube: 2 mm (int.Ø); 3 mm (ext.Ø)
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $940.76
    $940.76
    Log in for availability
    Each
    Each - $940.76
    $940.76
    Case of 4
       $940.76

    Each
       $940.76

    Supplier Number: C10333-DZC050B
  • 5 L Flasks
    90 cm (External)
    34 cm (Internal)
    External liquid transfer tube: 3.2 mm (int.Ø); 6.4 mm (ext.Ø)
    Internal liquid transfer tube: 2 mm (int.Ø); 3 mm (ext.Ø)
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $940.76
    $940.76
    Log in for availability
    Each
    Each - $940.76
    $940.76
    Case of 4
       $940.76

    Each
       $940.76

    Supplier Number: C10431-AZC090B
  • 5 L Flasks
    90 cm (External)
    34 cm (Internal)
    External liquid transfer tube: 6.4 mm (int.Ø); 11.1 mm (ext.Ø)
    Internal liquid transfer tube: 2 mm (int.Ø); 3 mm (ext.Ø)
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $1,112.88
    $1,112.88
    Log in for availability
    Each
    Each - $1,112.88
    $1,112.88
    Case of 4
       $1,112.88

    Each
       $1,112.88

    Supplier Number: C10433-DZC090B
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.