Aphidicolin, white ≥98% (by TLC), solid, Sigma-Aldrich®
1 mg
Supplier: MilliporeSigma
Supplier #: 178273-1MG
Aphidicolin, white ≥98% (by TLC), solid, Sigma-Aldrich®
Catalog # CA80055-804
Supplier: MilliporeSigma
CAS Number: 38966-21-1
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
A tetracyclic diterpene antibiotic
Cell synchronization agent. Blocks the cell cycle at early S-phase. Specific inhibitor of DNA polymerase [alpha] and [delta] in eukaryotic cells and in some viruses. Potentiates apoptosis induced by arabinosyl nucleosides in leukemia cell lines. Also induces apoptosis in HeLa S3 cells, but inhibits vincristine-induced apoptosis in the p53-negative human prostate cancer cell line PC-3.
Soluble in DMSO, EtOH, or MeOH
Synonyms: (3R,4R,4aR,6aS,8R,9R,11aS,11bS)-4,9-Bis(hydroxymethyl)-4,11b-dimethyltetradecahydro-8,11a-methanocyclohepta[a]naphthalene-3,9-diol
- CAS Number:38966-21-1
- Formula:C₂₀H₃₄O₄
- MDL Number:MFCD00083214
- Molecular Weight:338.488 g/mol
Documents
Specifications
Specifications
- Size:1 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



