ContactUsLinkComponent
Adjustable Slide Truck, Metro™
Catalog # CA82002-468
Supplier:  Metropolitan Wire
undefined
Adjustable Slide Truck, Metro™
Catalog # CA82002-468
Supplier:  Metropolitan Wire
Supplier Number:  APT1C-5M
Specifications
  • Description:
    Adjustable Slide Truck
  • Cat. no.:
    CA82002-468
  • Shipping Weight:
    54 lbs.
  • Supplier No.:
    APT1C-5M
Specifications
About this item
Chrome-plated truck is designed to store and transport carriers and tote boxes
Unit is adjustable in width from 52.4 to 74.6cm (205/8 to 293/8"). Use a socket wrench to easily adjust the width. The addition of an optional grounding bracket and cable (sold separately) will make this unit conductive.
Ordering information: Truck is supplied with four 12.7cm (5") resilient tread casters (two brake and two swivel). For Olympic™ tote boxes to use with the adjustable slide truck, see 22221-298 and 82002-506 series, or contact your Avantor representative.