About this item
Plasticrit Plastic Hematocrit Tubes provide similar capillary action to that of glass hematocrit tubes but eliminate the possibility of contamination and injury due to breakage.
- Polycarbonate Construction
- No Risk of Contamination or Injury Due to Breakage
- Length: 75 mm, 3"
- Tubes Comply with FDA, NIOSH, CDC, and OSHA Safety Recommendations
They exhibit no compromise in accuracy.
These 75 mm (3") tubes are made of polycarbonate.