Orders LinkContactUsLinkComponent
Capillary Tubes, Drummond
Capillary Tubes, Drummond
Capillary Tubes
Catalog # 13450-216
Supplier:   Drummond
Supplier #:  1-000-800/12
Capillary Tubes, Drummond
Catalog # 13450-216
Supplier:  Drummond
Capillary Tubes, Drummond
Capillary Tubes, Drummond
Capillary Tubes
Price
$28.16
New Customer Promotional Price
Log in for availability
Pack of 100 Case of 1200
Product Details & Documents
Glass capillary tubes for coagulation, hemoglobin, prothrombin, blood grouping and typing.
  • Capacity 9.4 µL
  • Length 7.5cm
  • Internal Diameter 0.4-0.6mm.
 : Packaged in vials of 100 tubes.
Specifications
  • Description:
    Capillary Tubes