ContactUsLinkComponent
408-MK Series Face Shields, OP-D-OP
408-MK Series Face Shields, OP-D-OP
Catalog # 33000-471
Supplier:  OP-D-OP
408-MK Series Face Shields, OP-D-OP
Catalog # 33000-471
Supplier:  OP-D-OP
Supplier Number:  408WT
Specifications
  • Description:
    408-MK Series Face Shields
  • Cat. no.:
    33000-471
  • Supplier No.:
    408WT
Specifications
About this item
Face shields are designed for use in emergency rooms, critical care, home care, dialysis, or any application requiring full face protection for 30 minutes or less.
  • The cordless visors are based on size medium (9 cm)
  • Fit over glasses and replace goggles and masks
  • Extremely durable polycarbonate shields are reusable and replaceable
Visor color: White
Ordering information: Each face shield kit includes one visor and two face shields. Additional visor colors and larger sizes are available. Contact your Avantor sales representative for more information.