CellPro™ NZY Broth, Molecular Biology Grade, Sterile Filtered
Catalog # 104029-016
Supplier: Alkali Scientific
CAS Number:
Specifications
- Media Format:Liquid
- Description:CellPro™ NZY broth, PC
- Size:500 ml
- Cat. no.:104029-016
Specifications
About this item
Prepare nutrient-rich broths for bacterial culture with Alkali Scientific CellPro™ NZY Broth, ensured sterile filtered and meeting molecular biology grade standards.
- Sterile
- Filtered
CellPro™ NZY Broth is a nutrient-rich growth medium designed for the cultivation of Escherichia coli strains. With its formulation of casein hydrolysate, yeast extract, and sodium chloride, this media supports rapid bacterial growth and high plasmid yield, making it ideal for molecular cloning and protein expression experiments. Through meticulous sterile filtration, CellPro™ NZY Broth ensures purity and consistency, empowering researchers with reliable results in molecular biology workflows.