ContactUsLinkComponent
Sample Loop, for 1290 Infinity II Series Preparative Open-Bed ALS/Collector G7158B
Sample Loop, for 1290 Infinity II Series Preparative Open-Bed ALS/Collector G7158B
Catalog # AG5068-0336
Sample Loop, for 1290 Infinity II Series Preparative Open-Bed ALS/Collector G7158B
Catalog # AG5068-0336
Supplier Number:  5068-0336
Specifications
  • Ø int.:
    ¹/₈"
  • Capacity:
    20 ml
  • Material:
    Stainless steel
  • Cat. no.:
    AG5068-0336