Orders LinkContactUsLinkComponent
Cell Culture Inserts
Supplier:   NEST SCIENTIFIC
Cell Culture Inserts
Supplier:  NEST SCIENTIFIC
Certificates
Cell Culture Inserts
47 Options Available Below
Product Details & Documents
Cell culture plates are made of high clarity, 100% virgin polystyrene.
  • Sterilized by E-beam, SAL=10⁻⁶
  • Vacuum plasma tissue culture treatment
  • Non-pyrogenic, DNase/RNase free
  • A rich selection of matching plates: 6-, 12-, 24-well
  • Innovative edge design for convenient sample loading
  • Low protein binding to ensure accurate results
Passed the USP VI toxicity test. Compatible with most solvents used for fixing and staining.
Product Family Options
PC membrane
Product Information
  • Well volume
    Ø
    Culture area
    Density
    Membrane
    Pore size
    Surface treatment
    Packaging
    Availability
    Price
  • 77508-640
    6 Cell culture inserts and 6 well plate, opaque
    6 Cell culture inserts and 6 well plate, opaque
    2.6 ml
    24 mm
    4.67 cm²
    1×10⁸
    PC membrane
    0.4 μm
    TC treated
    6/pack, 60/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $255.49
    $255.49
    Log in for availability
    Each
    Each - $255.49
    $255.49
    Case of 24
       $255.49
    Each
       $255.49
    Supplier Number: 723101
  • 77508-642
    6 Cell culture inserts and 6 well plate, opaque
    6 Cell culture inserts and 6 well plate, opaque
    2.6 ml
    24 mm
    4.67 cm²
    1×10⁸
    PC membrane
    0.4 μm
    Non-treated
    6/pack, 60/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $255.49
    $255.49
    Log in for availability
    Each
    Each - $255.49
    $255.49
    Case of 24
       $255.49
    Each
       $255.49
    Supplier Number: 723111
  • 77508-632
    6 Cell culture inserts and 6 well plate, translucent
    6 Cell culture inserts and 6 well plate, translucent
    2.6 ml
    24 mm
    4.67 cm²
    2×10⁶
    PC membrane
    3.0 μm
    TC treated
    6/pack, 60/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $255.49
    $255.49
    Log in for availability
    Each
    Each - $255.49
    $255.49
    Case of 24
       $255.49
    Each
       $255.49
    Supplier Number: 723001
  • 77508-634
    6 Cell culture inserts and 6 well plate, translucent
    6 Cell culture inserts and 6 well plate, translucent
    2.6 ml
    24 mm
    4.67 cm²
    2×10⁶
    PC membrane
    3.0 μm
    Non-treated
    6/pack, 60/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $255.49
    $255.49
    Log in for availability
    Each
    Each - $255.49
    $255.49
    Case of 24
       $255.49
    Each
       $255.49
    Supplier Number: 723011
  • 77508-648
    6 Cell culture inserts and 6 well plate, translucent
    6 Cell culture inserts and 6 well plate, translucent
    2.6 ml
    24 mm
    4.67 cm²
    1×10⁵
    PC membrane
    8.0 μm
    TC treated
    6/pack, 60/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $255.49
    $255.49
    Log in for availability
    Each
    Each - $255.49
    $255.49
    Case of 24
       $255.49
    Each
       $255.49
    Supplier Number: 723301
  • 77508-650
    6 Cell culture inserts and 6 well plate, translucent
    6 Cell culture inserts and 6 well plate, translucent
    2.6 ml
    24 mm
    4.67 cm²
    1×10⁵
    PC membrane
    8.0 μm
    Non-treated
    6/pack, 60/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $255.49
    $255.49
    Log in for availability
    Each
    Each - $255.49
    $255.49
    Case of 24
       $255.49
    Each
       $255.49
    Supplier Number: 723311
  • 77508-668
    12 Cell culture inserts and 12 well plate, opaque
    12 Cell culture inserts and 12 well plate, opaque
    1.5 ml
    12 mm
    1.12 cm²
    1×10⁸
    PC membrane
    0.4 μm
    PC treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $313.71
    $313.71
    Log in for availability
    Each
    Each - $313.71
    $313.71
    Case of 48
       $313.71
    Each
       $313.71
    Supplier Number: 724101
  • 77508-670
    12 Cell culture inserts and 12 well plate, opaque
    12 Cell culture inserts and 12 well plate, opaque
    1.5 ml
    12 mm
    1.12 cm²
    1×10⁸
    PC membrane
    0.4 μm
    Non-treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 724111
  • 77508-660
    12 Cell culture inserts and 12 well plate, translucent
    12 Cell culture inserts and 12 well plate, translucent
    1.5 ml
    12 mm
    1.12 cm²
    2×10⁶
    PC membrane
    3.0 μm
    PC treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $313.71
    $313.71
    Log in for availability
    Each
    Each - $313.71
    $313.71
    Case of 48
       $313.71
    Each
       $313.71
    Supplier Number: 724001
  • 77508-662
    12 Cell culture inserts and 12 well plate, translucent
    12 Cell culture inserts and 12 well plate, translucent
    1.5 ml
    12 mm
    1.12 cm²
    2×10⁶
    PC membrane
    3.0 μm
    Non-treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 724011
  • 77508-676
    12 Cell culture inserts and 12 well plate, translucent
    12 Cell culture inserts and 12 well plate, translucent
    1.5 ml
    12 mm
    1.12 cm²
    4×10⁵
    PC membrane
    5.0 μm
    PC treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 724201
  • 77508-678
    12 Cell culture inserts and 12 well plate, translucent
    12 Cell culture inserts and 12 well plate, translucent
    1.5 ml
    12 mm
    1.12 cm²
    4×10⁵
    PC membrane
    5.0 μm
    Non-treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 724211
  • 77508-680
    12 Cell culture inserts and 12 well plate, translucent
    12 Cell culture inserts and 12 well plate, translucent
    1.5 ml
    12 mm
    1.12 cm²
    1×10⁵
    PC membrane
    8.0 μm
    PC treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 724301
  • 77508-682
    12 Cell culture inserts and 12 well plate, translucent
    12 Cell culture inserts and 12 well plate, translucent
    1.5 ml
    12 mm
    1.12 cm²
    1×10⁵
    PC membrane
    8.0 μm
    Non-treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $313.71
    $313.71
    Log in for availability
    Each
    Each - $313.71
    $313.71
    Case of 48
       $313.71
    Each
       $313.71
    Supplier Number: 724311
  • 77508-700
    12 Cell culture inserts and 24 well plate, opaque
    12 Cell culture inserts and 24 well plate, opaque
    0.6 ml
    6.5 mm
    0.33 cm²
    1×10⁸
    PC membrane
    0.4 μm
    PC treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 725101
  • 77508-702
    12 Cell culture inserts and 24 well plate, opaque
    12 Cell culture inserts and 24 well plate, opaque
    0.6 ml
    6.5 mm
    0.33 cm²
    1×10⁸
    PC membrane
    0.4 μm
    Non-treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 725111
  • 77508-692
    12 Cell culture inserts and 24 well plate, translucent
    12 Cell culture inserts and 24 well plate, translucent
    0.6 ml
    6.5 mm
    0.33 cm²
    2×10⁶
    PC membrane
    3.0 μm
    PC treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 725001
  • 77508-694
    12 Cell culture inserts and 24 well plate, translucent
    12 Cell culture inserts and 24 well plate, translucent
    0.6 ml
    6.5 mm
    0.33 cm²
    2×10⁶
    PC membrane
    3.0 μm
    Non-treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 725011
  • 77508-708
    12 Cell culture inserts and 24 well plate, translucent
    12 Cell culture inserts and 24 well plate, translucent
    0.6 ml
    6.5 mm
    0.33 cm²
    4×10⁵
    PC membrane
    5.0 μm
    PC treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 725201
  • 77508-710
    12 Cell culture inserts and 24 well plate, translucent
    12 Cell culture inserts and 24 well plate, translucent
    0.6 ml
    6.5 mm
    0.33 cm²
    4×10⁵
    PC membrane
    5.0 μm
    Non-treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 725211
  • 77508-712
    12 Cell culture inserts and 24 well plate, translucent
    12 Cell culture inserts and 24 well plate, translucent
    0.6 ml
    6.5 mm
    0.33 cm²
    1×10⁵
    PC membrane
    8.0 μm
    PC treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 725301
  • 77508-714
    12 Cell culture inserts and 24 well plate, translucent
    12 Cell culture inserts and 24 well plate, translucent
    0.6 ml
    6.5 mm
    0.33 cm²
    1×10⁵
    PC membrane
    8.0 μm
    Non-treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 725311
  • 77508-724
    Cell culture insert-dish, 100 mm, translucent
    Cell culture insert-dish, 100 mm, translucent
    13 ml
    75 mm
    44 cm²
    2×10⁶
    PC membrane
    3.0 μm
    PC treated
    1/pack, 10/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $616.10
    $616.10
    Log in for availability
    Each
    Each - $616.10
    $616.10
    Case of 10
       $616.10
    Each
       $616.10
    Supplier Number: 726001
PET membrane
Product Information
  • Well volume
    Ø
    Culture area
    Density
    Membrane
    Pore size
    Surface treatment
    Packaging
    Availability
    Price
  • 77508-644
    6 Cell culture inserts and 6 well plate, opaque
    6 Cell culture inserts and 6 well plate, opaque
    2.6 ml
    24 mm
    4.67 cm²
    1×10⁸
    PET membrane
    0.4 μm
    TC treated
    6/pack, 60/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $215.02
    $215.02
    Log in for availability
    Each
    Each - $215.02
    $215.02
    Case of 24
       $215.02
    Each
       $215.02
    Supplier Number: 723121
  • 77508-646
    6 Cell culture inserts and 6 well plate, opaque
    6 Cell culture inserts and 6 well plate, opaque
    2.6 ml
    24 mm
    4.67 cm²
    1×10⁸
    PET membrane
    0.4 μm
    Non-treated
    6/pack, 60/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $213.09
    $213.09
    Log in for availability
    Each
    Each - $213.09
    $213.09
    Case of 24
       $213.09
    Each
       $213.09
    Supplier Number: 723131
  • 77508-656
    6 Cell culture inserts and 6 well plate, transparent
    6 Cell culture inserts and 6 well plate, transparent
    2.6 ml
    24 mm
    4.67 cm²
    2×10⁷
    PET membrane
    1.0 μm
    TC treated
    6/pack, 60/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $213.09
    $213.09
    Log in for availability
    Each
    Each - $213.09
    $213.09
    Case of 24
       $213.09
    Each
       $213.09
    Supplier Number: 723421
  • 77508-658
    6 Cell culture inserts and 6 well plate, transparent
    6 Cell culture inserts and 6 well plate, transparent
    2.6 ml
    24 mm
    4.67 cm²
    2×10⁷
    PET membrane
    1.0 μm
    Non-treated
    6/pack, 60/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $213.09
    $213.09
    Log in for availability
    Each
    Each - $213.09
    $213.09
    Case of 24
       $213.09
    Each
       $213.09
    Supplier Number: 723431
  • 77508-636
    6 Cell culture inserts and 6 well plate, translucent
    6 Cell culture inserts and 6 well plate, translucent
    2.6 ml
    24 mm
    4.67 cm²
    2×10⁶
    PET membrane
    3.0 μm
    TC treated
    6/pack, 60/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $213.09
    $213.09
    Log in for availability
    Each
    Each - $213.09
    $213.09
    Case of 24
       $213.09
    Each
       $213.09
    Supplier Number: 723021
  • 77508-638
    6 Cell culture inserts and 6 well plate, translucent
    6 Cell culture inserts and 6 well plate, translucent
    2.6 ml
    24 mm
    4.67 cm²
    2×10⁶
    PET membrane
    3.0 μm
    Non-treated
    6/pack, 60/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $213.09
    $213.09
    Log in for availability
    Each
    Each - $213.09
    $213.09
    Case of 24
       $213.09
    Each
       $213.09
    Supplier Number: 723031
  • 77508-652
    6 Cell culture inserts and 6 well plate, transparent
    6 Cell culture inserts and 6 well plate, transparent
    2.6 ml
    24 mm
    4.67 cm²
    1×10⁵
    PET membrane
    8.0 μm
    TC treated
    6/pack, 60/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $215.02
    $215.02
    Log in for availability
    Each
    Each - $215.02
    $215.02
    Case of 24
       $215.02
    Each
       $215.02
    Supplier Number: 723321
  • 77508-654
    6 Cell culture inserts and 6 well plate, transparent
    6 Cell culture inserts and 6 well plate, transparent
    2.6 ml
    24 mm
    4.67 cm²
    1×10⁵
    PET membrane
    8.0 μm
    Non-treated
    6/pack, 60/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $215.02
    $215.02
    Log in for availability
    Each
    Each - $215.02
    $215.02
    Case of 24
       $215.02
    Each
       $215.02
    Supplier Number: 723331
  • 77508-672
    12 Cell culture inserts and 12 well plate, opaque
    12 Cell culture inserts and 12 well plate, opaque
    1.5 ml
    12 mm
    1.12 cm²
    1×10⁸
    PET membrane
    0.4 μm
    TC treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 724121
  • 77508-674
    12 Cell culture inserts and 12 well plate, opaque
    12 Cell culture inserts and 12 well plate, opaque
    1.5 ml
    12 mm
    1.12 cm²
    1×10⁸
    PET membrane
    0.4 μm
    Non-treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $313.71
    $313.71
    Log in for availability
    Each
    Each - $313.71
    $313.71
    Case of 48
       $313.71
    Each
       $313.71
    Supplier Number: 724131
  • 77508-688
    12 Cell culture inserts and 12 well plate, transparent
    12 Cell culture inserts and 12 well plate, transparent
    1.5 ml
    12 mm
    1.12 cm²
    2×10⁷
    PET membrane
    1.0 μm
    TC treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 724421
  • 77508-690
    12 Cell culture inserts and 12 well plate, transparent
    12 Cell culture inserts and 12 well plate, transparent
    1.5 ml
    12 mm
    1.12 cm²
    2×10⁷
    PET membrane
    1.0 μm
    Non-treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 724431
  • 77508-664
    12 Cell culture inserts and 12 well plate, translucent
    12 Cell culture inserts and 12 well plate, translucent
    1.5 ml
    12 mm
    1.12 cm²
    2×10⁶
    PET membrane
    3.0 μm
    TC treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 724021
  • 77508-666
    12 Cell culture inserts and 12 well plate, translucent
    12 Cell culture inserts and 12 well plate, translucent
    1.5 ml
    12 mm
    1.12 cm²
    2×10⁶
    PET membrane
    3.0 μm
    Non-treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 724031
  • 77508-684
    12 Cell culture inserts and 12 well plate, transparent
    12 Cell culture inserts and 12 well plate, transparent
    1.5 ml
    12 mm
    1.12 cm²
    1×10⁵
    PET membrane
    8.0 μm
    TC treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 724321
  • 77508-686
    12 Cell culture inserts and 12 well plate, transparent
    12 Cell culture inserts and 12 well plate, transparent
    1.5 ml
    12 mm
    1.12 cm²
    1×10⁵
    PET membrane
    8.0 μm
    Non-treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $313.71
    $313.71
    Log in for availability
    Each
    Each - $313.71
    $313.71
    Case of 48
       $313.71
    Each
       $313.71
    Supplier Number: 724331
  • 77508-704
    12 Cell culture inserts and 24 well plate, opaque
    12 Cell culture inserts and 24 well plate, opaque
    0.6 ml
    6.5 mm
    0.33 cm²
    1×10⁸
    PET membrane
    0.4 μm
    TC treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 725121
  • 77508-706
    12 Cell culture inserts and 24 well plate, opaque
    12 Cell culture inserts and 24 well plate, opaque
    0.6 ml
    6.5 mm
    0.33 cm²
    1×10⁸
    PET membrane
    0.4 μm
    Non-treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 725131
  • 77508-720
    12 Cell culture inserts and 24 well plate, transparent
    12 Cell culture inserts and 24 well plate, transparent
    0.6 ml
    6.5 mm
    0.33 cm²
    2×10⁷
    PET membrane
    1.0 μm
    TC treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 725421
  • 77508-722
    12 Cell culture inserts and 24 well plate, transparent
    12 Cell culture inserts and 24 well plate, transparent
    0.6 ml
    6.5 mm
    0.33 cm²
    2×10⁷
    PET membrane
    1.0 μm
    Non-treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 725431
  • 77508-696
    12 Cell culture inserts and 24 well plate, translucent
    12 Cell culture inserts and 24 well plate, translucent
    0.6 ml
    6.5 mm
    0.33 cm²
    2×10⁶
    PET membrane
    3.0 μm
    TC treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 725021
  • 77508-698
    12 Cell culture inserts and 24 well plate, translucent
    12 Cell culture inserts and 24 well plate, translucent
    0.6 ml
    6.5 mm
    0.33 cm²
    2×10⁶
    PET membrane
    3.0 μm
    Non-treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 725031
  • 77508-716
    12 Cell culture inserts and 24 well plate, transparent
    12 Cell culture inserts and 24 well plate, transparent
    0.6 ml
    6.5 mm
    0.33 cm²
    1×10⁵
    PET membrane
    8.0 μm
    TC treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $313.71
    $313.71
    Log in for availability
    Each
    Each - $313.71
    $313.71
    Case of 48
       $313.71
    Each
       $313.71
    Supplier Number: 725321
  • 77508-718
    12 Cell culture inserts and 24 well plate, transparent
    12 Cell culture inserts and 24 well plate, transparent
    0.6 ml
    6.5 mm
    0.33 cm²
    1×10⁵
    PET membrane
    8.0 μm
    Non-treated
    12/pack, 120/case
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $310.89
    $310.89
    Log in for availability
    Each
    Each - $310.89
    $310.89
    Case of 48
       $310.89
    Each
       $310.89
    Supplier Number: 725331