Pipette Tips, 10 ml
Catalog # 104027-626
Supplier: Alkali Scientific
Specifications
- Volume:1000 - 10000 µl
- Packed:150/pack
- Cat. no.:104027-626
- Sterile:+
Specifications
About this item
Handle larger volumes with ease using Alkali Scientific™ 10 ml pipette tips, designed for accurate and efficient liquid handling.
- Ultra-clear polypropylene
Enhance your liquid handling efficiency with Alkali Scientific™ 10 ml pipette tips. Designed for use with compatible pipets, these tips are meticulously crafted from high-quality materials to ensure accurate and precise liquid transfers. Their ergonomic design and secure fit provide ease of use and confidence in pipetting tasks, making them essential accessories for laboratories and research facilities. Trust Alkali Scientific™ 10 ml pipette tips for reliable performance in your daily laboratory operations.