Conductivity Standard, 500000 µS/cm (500 mS/cm) at 25 °C
RCR2249500500A
New Product
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Pack type:500 ml plastic
- Environmentally Preferable:
- Reference Temperature:25 °C
- Cat. no.:RCR2249500500A
- Conductivity:500000 µS/cm
- Certified traceable to NIST Standard Reference Materials
- Tested in ISO 17025 Accredited Laboratories
Calibrate with confidence using RICCA Conductivity Standards. Suitalbe for use with any probe. Tested to the tightest specifications in the industry, you can count on RICCA conductivity buffers to produce accurate and reproducible readings with every pour.