Order Entry
Canada
ContactUsLinkComponent
Conductivity Standard, 400000 µS/cm (400 mS/cm) at 25 °C
Conductivity Standard, 400000 µS/cm (400 mS/cm) at 25 °C
Catalog # RCR2249400500A
Supplier:  Ricca Chemical
CAS Number:  
undefined
Conductivity Standard, 400000 µS/cm (400 mS/cm) at 25 °C
Catalog # RCR2249400500A
Supplier:  Ricca Chemical
Supplier Number:  R2249400-500A
CAS Number:  

Specifications

  • Pack type:
    500 ml plastic
  • Reference Temperature:
    25 °C
  • Cat. no.:
    RCR2249400500A
  • Conductivity:
    400000 µS/cm

Specifications

About this item

  • Certified traceable to NIST Standard Reference Materials
  • Tested in ISO 17025 Accredited Laboratories

Calibrate with confidence using RICCA Conductivity Standards. Suitalbe for use with any probe. Tested to the tightest specifications in the industry, you can count on RICCA conductivity buffers to produce accurate and reproducible readings with every pour.