Specifications
- Description:Blood Dispensers
- Cat. no.:CA73000-077
- Supplier No.:101
Specifications
About this item
Designed to minimize airborne pathogens by eliminating the need to remove the stopper from the blood tube
Designed to minimize airborne pathogens by eliminating the need to remove the stopper from the blood tube
The dispensers feature a cannula that inserts easily through the rubber stopper of a blood collection tube. When the tube is inverted and the dispenser pressed against a slide, it dispenses a drop ideally sized for making a blood smear. Compatible with BD Vacutainer® tubes with BD Hemogard™ closures. Nonsterile.