
Sodium dodecyl sulfate (SDS) 10%, CellPro™
Catalog # 104027-148
Supplier: Alkali Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:1 L x 12/cs
- Cat. no.:104027-148
Specifications
About this item
:
:288.38 g/mol
UN:3082
:GHS09
CAS Number:151-21-3
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175