Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
di-Sodium hydrogen phosphate
Disodium hydrogen phosphate solution 1M, CellPro™
Disodium hydrogen phosphate solution 1M, CellPro™
104026-634
:
Alkali Scientific
:
Disodium hydrogen phosphate solution 1M, CellPro™
104026-634
:
Alkali Scientific
:
DS0327
:
Pack type:
Plastic bottle
Size:
1 L
Cat. no.:
104026-634
:
:
141.96 g/mol
:
3248
:
7558-79-4
:
Na₂HPO₄
:
MFCD00003496