Order Entry
Canada
ContactUsLinkComponent
Kanamycin solution 10X 100 µg/ml, CellPro™
Kanamycin solution 10X 100 µg/ml, CellPro™
Catalog # 104027-512
Supplier:  Alkali Scientific
CAS Number:  
undefined
Kanamycin solution 10X 100 µg/ml, CellPro™
Catalog # 104027-512
Supplier:  Alkali Scientific
Supplier Number:  DS0201
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    100 ml
  • Cat. no.:
    104027-512

Specifications

About this item

 :
UN:3248
CAS Number:59-01-8
MDL Number:MFCD00070289