IPTG (isopropyl-β-D-thiogalactopyranoside) solution 5mM, CellPro™
Catalog # 104027-094
Supplier: Alkali Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:10 ml
- Cat. no.:104027-094
Specifications
About this item
:
:238.31 g/mol
UN:3248
CAS Number:367-93-1
Formula:C₉H₁₈O₅S
MDL Number:MFCD00063273