Orders LinkContactUsLinkComponent
Microscope Cover Glasses
Microscope Cover Glasses
Supplier:   NEST SCIENTIFIC
Microscope Cover Glasses
Supplier:  NEST SCIENTIFIC
Certificates
Microscope Cover Glasses
5 Options Available Below
Product Details & Documents
Microscope cover glasses are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials.
  • Resistance to organic solvent
  • Autoclavable
  • Can be used in confocal experiment
  • High quality glass, non-cytotoxic
  • Thickness: 0.16 to 0.19 mm(±0.02 mm)
Product Family Options
Product Information
  • Packed
    Ø
    Material
    Availability
    Price
  • 77508-940
    24 well cell culture plate
    24 well cell culture plate
    100/pack
    14 mm
    Glass
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $120.70
    $120.70
    Log in for availability
    Each
    Each - $120.70
    $120.70
    Case of 100
       $120.70
    Each
       $120.70
    Supplier Number: 801010
  • 77508-934
    24 well cell culture plate
    24 well cell culture plate
    100/pack
    15 mm
    Glass
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $120.70
    $120.70
    Log in for availability
    Each
    Each - $120.70
    $120.70
    Case of 100
       $120.70
    Each
       $120.70
    Supplier Number: 801007
  • 77508-942
    12 well cell culture plate
    12 well cell culture plate
    100/pack
    18 mm
    Glass
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $120.70
    $120.70
    Log in for availability
    Each
    Each - $120.70
    $120.70
    Case of 100
       $120.70
    Each
       $120.70
    Supplier Number: 801011
  • 77508-936
    12 well cell culture plate
    12 well cell culture plate
    100/pack
    20 mm
    Glass
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $120.70
    $120.70
    Log in for availability
    Each
    Each - $120.70
    $120.70
    Case of 100
       $120.70
    Each
       $120.70
    Supplier Number: 801008
  • 77508-938
    6 well cell culture plate
    6 well cell culture plate
    100/pack
    25 mm
    Glass
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $120.70
    $120.70
    Log in for availability
    Each
    Each - $120.70
    $120.70
    Case of 100
       $120.70
    Each
       $120.70
    Supplier Number: 801009