L-Shaped Spreader
L-Shaped spreader, Blue, 200/pack, 2000/case
Catalog # 77508-584
Supplier: NEST SCIENTIFIC
: 711001
Jump to:
Product Details & Documents
Using a grinding tool with high precision in combination with an ergonomic design, the efficient sterile L-Spreader specialized for Asian scientific researchers is developed and manufactured to effectively solve problems such as contamination or uneven application of spread plate in bio-labs, which helps to make domestic biological testing
practitioners more efficient and simplify their tedious work during biological experiments.
- Sterile L-Spreader, using medical PS
- EO Sterilized
- Smooth and flat surface that solves problems such as contamination or uneven application of spread plate in bio-labs
- Length of handle: 144 mm, diagram of spreading: 33 mm
- Ideal for spreading cells or bacteria evenly in the culture dish
Suitable for 3.5, 6, 9 and 10 cm petri dishes and more.
Specifications
- Color:Blue
- Description:L-Shaped spreader
- Packaging:200/pack, 2000/case



