Pierce™ In-Solution Tryptic Digestion and Guanidination Kit, Thermo Scientific
CAPI89895
:
- Description:In-Solution Tryptic Digestion and Guanidation Kit
- Cat. no.:CAPI89895
- No. of Reactions:90
- Supplier No.:89895
Pierce™ In-Solution Tryptic Digestion and Guanidination Kit contains proteomics grade trypsin, buffers and reagents to alkylate and digest proteins and then guanidinate the peptide fragments for mass spectrometry.
- Optimized trypsin digestion producing complete digests of protein ranging from 0.025 to 10 µg
- Optimized guanidination where reaction produces little or no N-terminal modification
- Protein can be digested and guanidinated all in a day
Accurate identification of proteins and analysis of post-translational modifications by mass spectrometry require accurate and complete protein digestion and peptide modification.