Order Entry
Canada
Orders LinkContactUsLinkComponent
Storage Bottles, Square, Closed System Solution

Storage Bottles, Square, Closed System Solution

Storage Bottles, Square, Closed System Solution
Supplier:  NEST SCIENTIFIC
Certificates
Storage Bottles, Square, Closed System Solution

17 Options Available Below

Product Details & Documents

The closed system storage bottles collection provides an aseptic liquid infusion solution for pharmaceuticals, biotechnology industry and laboratories.

  • Various tubing specifications
  • The injection-molded transfer cap is highly compatible with other major brands
  • The bottle body is made of PETG, which has high transparency, mechanical strength, low-temperature resistance, and UV resistance, making it suitable for observation and transportation
  • The unique external thick collar design of the bottle neck secures a sealed environment for experiments
  • Non-cytotoxic, non-pyrogenic, no animal-derived components
  • Ten bottles per case
  • Pore size: 0.2 µm
  • Membrane are: 4.5 cm²

A standard Luer head is used in the joint of the system, enabling the operation more convenient. Also, the TPE material of tubes is in line with the requirements for industrial pharmaceutical enterprises, and is validated by a certified third party in accordance with BPOG Guide for Evaluating Extractables and Leachables.

Documents

Product Family Options

Bi-directional transfer cap bottles

Product Information

  • Capacity
    Material
    Tubing int.Ø
    Tubing material
    Tubing length
    Tubing ext.Ø
    Sterility
    Availability
    Price
  • 77509-124
    Closed system storage bottle, male CPC connector with female sealing cap
    Closed system storage bottle, male CPC connector with female sealing cap
    250 ml
    PETG
    ¹/₄"
    TPE
    60 cm
    ³/₈"
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $3,020.40
    $3,020.40
    Log in for availability
    Each
    Each - $3,020.40
    $3,020.40
    Case of 10
       $3,020.40

    Each
       $3,020.40

    Supplier Number: C50921-BBB060A
  • 77509-128
    Closed system storage bottle, female luer Lock connector with luer plug
    Closed system storage bottle, female luer Lock connector with luer plug
    250 ml
    PETG
    ¹/₄"
    TPE
    60 cm
    ³/₈"
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $1,739.57
    $1,739.57
    Log in for availability
    Each
    Each - $1,739.57
    $1,739.57
    Case of 10
       $1,739.57

    Each
       $1,739.57

    Supplier Number: C50921-BHB060A
  • 77509-132
    Closed system storage bottle, with bi-directional transfer cap
    Closed system storage bottle, with bi-directional transfer cap
    250 ml
    PETG
    ¹/₄"
    TPE
    60 cm
    ⁷/₁₆
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $2,083.83
    $2,083.83
    Log in for availability
    Each
    Each - $2,083.83
    $2,083.83
    Case of 10
       $2,083.83

    Each
       $2,083.83

    Supplier Number: C50921-DZB060A
  • 77509-134
    Closed system storage bottle, male CPC connector with female sealing cap
    Closed system storage bottle, male CPC connector with female sealing cap
    250 ml
    PETG
    ¹/₈"
    TPE
    60 cm
    ¹/₄"
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $2,827.57
    $2,827.57
    Log in for availability
    Each
    Each - $2,827.57
    $2,827.57
    Case of 10
       $2,827.57

    Each
       $2,827.57

    Supplier Number: C50922-AAB060A
  • 77509-138
    Closed system storage bottle, female luer Lock connector with luer plug
    Closed system storage bottle, female luer Lock connector with luer plug
    250 ml
    PETG
    ¹/₈"
    TPE
    60 cm
    ¹/₄"
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $1,477.32
    $1,477.32
    Log in for availability
    Each
    Each - $1,477.32
    $1,477.32
    Case of 10
       $1,477.32

    Each
       $1,477.32

    Supplier Number: C50922-AGB060A
  • 77509-142
    Closed system storage bottle, male CPC connector with female sealing cap
    Closed system storage bottle, male CPC connector with female sealing cap
    500 ml
    PETG
    ¹/₄"
    TPE
    60 cm
    ³/₈"
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $3,054.89
    $3,054.89
    Log in for availability
    Each
    Each - $3,054.89
    $3,054.89
    Case of 10
       $3,054.89

    Each
       $3,054.89

    Supplier Number: C51021-BBB060A
  • 77509-146
    Closed system storage bottle, female luer Lock connector with luer plug
    Closed system storage bottle, female luer Lock connector with luer plug
    500 ml
    PETG
    ¹/₄"
    TPE
    60 cm
    ³/₈"
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $1,790.06
    $1,790.06
    Log in for availability
    Each
    Each - $1,790.06
    $1,790.06
    Case of 10
       $1,790.06

    Each
       $1,790.06

    Supplier Number: C51021-BHB060A
  • 77509-150
    Closed system storage bottle, with bi-directional transfer cap
    Closed system storage bottle, with bi-directional transfer cap
    500 ml
    PETG
    ¹/₄"
    TPE
    60 cm
    ⁷/₁₆
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $2,118.34
    $2,118.34
    Log in for availability
    Each
    Each - $2,118.34
    $2,118.34
    Case of 10
       $2,118.34

    Each
       $2,118.34

    Supplier Number: C51021-DZB060A
  • 77509-152
    Closed system storage bottle, male CPC connector with female sealing cap
    Closed system storage bottle, male CPC connector with female sealing cap
    500 ml
    PETG
    ¹/₈"
    TPE
    60 cm
    ¹/₄"
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $2,862.08
    $2,862.08
    Log in for availability
    Each
    Each - $2,862.08
    $2,862.08
    Case of 10
       $2,862.08

    Each
       $2,862.08

    Supplier Number: C51022-AAB060A
  • 77509-156
    Closed system storage bottle, female luer Lock connector with luer plug
    Closed system storage bottle, female luer Lock connector with luer plug
    500 ml
    PETG
    ¹/₈"
    TPE
    60 cm
    ¹/₄"
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $1,512.11
    $1,512.11
    Log in for availability
    Each
    Each - $1,512.11
    $1,512.11
    Case of 10
       $1,512.11

    Each
       $1,512.11

    Supplier Number: C51022-AGB060A
  • 77509-160
    Closed system storage bottle, male CPC connector with female sealing cap
    Closed system storage bottle, male CPC connector with female sealing cap
    500 ml
    PETG
    ¹/₄"
    TPE
    60 cm
    ³/₈"
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $3,123.72
    $3,123.72
    Log in for availability
    Each
    Each - $3,123.72
    $3,123.72
    Case of 10
       $3,123.72

    Each
       $3,123.72

    Supplier Number: C51121-BBB060A
  • 77509-164
    Closed system storage bottle, female luer Lock connector with luer plug
    Closed system storage bottle, female luer Lock connector with luer plug
    1000 ml
    PETG
    ¹/₄"
    TPE
    60 cm
    ³/₈"
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $1,842.79
    $1,842.79
    Log in for availability
    Each
    Each - $1,842.79
    $1,842.79
    Case of 10
       $1,842.79

    Each
       $1,842.79

    Supplier Number: C51121-BHB060A
  • 77509-168
    Closed system storage bottle, with bi-directional transfer cap
    Closed system storage bottle, with bi-directional transfer cap
    1000 ml
    PETG
    ¹/₄"
    TPE
    60 cm
    ⁷/₁₆
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $2,187.17
    $2,187.17
    Log in for availability
    Each
    Each - $2,187.17
    $2,187.17
    Case of 10
       $2,187.17

    Each
       $2,187.17

    Supplier Number: C51121-DZB060A
  • 77509-170
    Closed system storage bottle, male CPC connector with female sealing cap
    Closed system storage bottle, male CPC connector with female sealing cap
    1000 ml
    PETG
    ¹/₈"
    TPE
    60 cm
    ¹/₄"
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $2,930.89
    $2,930.89
    Log in for availability
    Each
    Each - $2,930.89
    $2,930.89
    Case of 10
       $2,930.89

    Each
       $2,930.89

    Supplier Number: C51122-AAB060A
  • 77509-174
    Closed system storage bottle, male CPC connector with female sealing cap
    Closed system storage bottle, male CPC connector with female sealing cap
    1000 ml
    PETG
    ¹/₈"
    TPE
    60 cm
    ¹/₄"
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $1,567.32
    $1,567.32
    Log in for availability
    Each
    Each - $1,567.32
    $1,567.32
    Case of 10
       $1,567.32

    Each
       $1,567.32

    Supplier Number: C51122-AFB060A
  • 77509-176
    Closed system storage bottle, female luer lock connector with luer plug
    Closed system storage bottle, female luer lock connector with luer plug
    1000 ml
    PETG
    ¹/₈"
    TPE
    60 cm
    ¹/₄"
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $1,514.21
    $1,514.21
    Log in for availability
    Each
    Each - $1,514.21
    $1,514.21
    Case of 10
       $1,514.21

    Each
       $1,514.21

    Supplier Number: C51122-AGB060A

3-Port transfer cap bottle

Product Information

  • Capacity
    Material
    Tubing int.Ø
    Tubing material
    Tubing length
    Tubing ext.Ø
    Sterility
    Availability
    Price
  • 77509-180
    Closed system storage bottle, female luer lock connector with luer plug
    Closed system storage bottle, female luer lock connector with luer plug
    1000 ml
    PETG
    ¹/₈"
    TPE
    60 cm
    ¹/₄"
    Sterile
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $2,035.52
    $2,035.52
    Log in for availability
    Each
    Each - $2,035.52
    $2,035.52
    Case of 10
       $2,035.52

    Each
       $2,035.52

    Supplier Number: C511AB-AGB060A

Bi-directional transfer caps

Product Information

  • Catalog #
    Description
    For use with
    Availability
    Price
  • Bi-directional transfer cap
    500 ml Square storage bottle closed system solution
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $1,946.11
    $1,946.11
    Log in for availability
    Each
    Each - $1,946.11
    $1,946.11
    Case of 10
       $1,946.11

    Each
       $1,946.11

Product Information

  • Catalog #
    Description
    For use with
    Availability
    Price
  • Bi-directional transfer cap
    500 ml Square storage bottle closed system solution
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $1,670.65
    $1,670.65
    Log in for availability
    Each
    Each - $1,670.65
    $1,670.65
    Case of 10
       $1,670.65

    Each
       $1,670.65

Product Information

  • Catalog #
    Description
    For use with
    Availability
    Price
  • Bi-directional transfer cap
    1000 ml Square storage bottle closed system solution
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $3,227.03
    $3,227.03
    Log in for availability
    Each
    Each - $3,227.03
    $3,227.03
    Case of 10
       $3,227.03

    Each
       $3,227.03

Product Information

  • Catalog #
    Description
    For use with
    Availability
    Price
  • Bi-directional transfer cap
    1000 ml Square storage bottle closed system solution
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $1,946.11
    $1,946.11
    Log in for availability
    Each
    Each - $1,946.11
    $1,946.11
    Case of 10
       $1,946.11

    Each
       $1,946.11

Product Information

  • Catalog #
    Description
    For use with
    Availability
    Price
  • Bi-directional transfer cap
    250 ml Square storage bottle closed system solution
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $3,227.03
    $3,227.03
    Log in for availability
    Each
    Each - $3,227.03
    $3,227.03
    Case of 10
       $3,227.03

    Each
       $3,227.03

Product Information

  • Catalog #
    Description
    For use with
    Availability
    Price
  • Bi-directional transfer cap
    250 ml Square storage bottle closed system solution
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $1,946.11
    $1,946.11
    Log in for availability
    Each
    Each - $1,946.11
    $1,946.11
    Case of 10
       $1,946.11

    Each
       $1,946.11

Product Information

  • Catalog #
    Description
    For use with
    Availability
    Price
  • Bi-directional transfer cap
    250 ml Square storage bottle closed system solution
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $3,034.21
    $3,034.21
    Log in for availability
    Each
    Each - $3,034.21
    $3,034.21
    Case of 10
       $3,034.21

    Each
       $3,034.21

Product Information

  • Catalog #
    Description
    For use with
    Availability
    Price
  • Bi-directional transfer cap
    250 ml Square storage bottle closed system solution
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $1,670.65
    $1,670.65
    Log in for availability
    Each
    Each - $1,670.65
    $1,670.65
    Case of 10
       $1,670.65

    Each
       $1,670.65

Product Information

  • Catalog #
    Description
    For use with
    Availability
    Price
  • Bi-directional transfer cap
    500 ml Square storage bottle closed system solution
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $3,227.03
    $3,227.03
    Log in for availability
    Each
    Each - $3,227.03
    $3,227.03
    Case of 10
       $3,227.03

    Each
       $3,227.03

Product Information

  • Catalog #
    Description
    For use with
    Availability
    Price
  • Bi-directional transfer cap
    1000 ml Square storage bottle closed system solution
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $3,034.21
    $3,034.21
    Log in for availability
    Each
    Each - $3,034.21
    $3,034.21
    Case of 10
       $3,034.21

    Each
       $3,034.21

Product Information

  • Catalog #
    Description
    For use with
    Availability
    Price
  • Bi-directional transfer cap
    1000 ml Square storage bottle closed system solution
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $1,685.87
    $1,685.87
    Log in for availability
    Each
    Each - $1,685.87
    $1,685.87
    Case of 10
       $1,685.87

    Each
       $1,685.87

Product Information

  • Catalog #
    Description
    For use with
    Availability
    Price
  • Bi-directional transfer cap
    500 ml Square storage bottle closed system solution
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $3,034.21
    $3,034.21
    Log in for availability
    Each
    Each - $3,034.21
    $3,034.21
    Case of 10
       $3,034.21

    Each
       $3,034.21

3-Port transfer cap

Product Information

  • Catalog #
    Description
    For use with
    Availability
    Price
  • 3-Port transfer cap
    1000 ml Square storage bottle closed system solution
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $1,863.48
    $1,863.48
    Log in for availability
    Each
    Each - $1,863.48
    $1,863.48
    Case of 10
       $1,863.48

    Each
       $1,863.48