Order Entry
Canada
ContactUsLinkComponent
Phosphate Buffered Saline, 1X
Phosphate Buffered Saline, 1X
Catalog # 77510-382
Supplier:  RPI
CAS Number:  
undefined
Phosphate Buffered Saline, 1X
Catalog # 77510-382
Supplier:  RPI
Supplier Number:  P10450-36.0
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Phosphate buffered saline 1X sterile solution
  • Size:
    1 ml
  • Sterility:
    Sterile
  • Storage temperature:
    RT
  • Appearance:
    Clear
  • Cat. no.:
    77510-382
  • pH:
    7.25 - 7.55

Specifications

About this item

Ready to use sterile 1X solution, supplied in sterile pre-filled screw cap tubes. Phosphate buffered saline is a buffer solution commonly used in biological research. PBS has many uses because it is isotonic and non-toxic to most cells.

  • Sterility: Filtered through 0.22 µm filter; Autoclaved 15 min at 121 °C
  • Solubility: Water
Packaging: Each tube contains 1.0 ml PBS 1X sterile solution, packaged in a sealed white cardboard freezer box with a 6×6 divider. PBS is supplied in 4.5 ml screw cap tubes that are 3.5" high (cap on). Each case holds 36 pre-filled tubes.
Caution: Research or further manufacturing use only, not for food or drug use.