Nunc® Thermanox™ Tissue Culture Coverslips, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Thermanox™ coverslips provide excellent microscopic analysis with minimal cell disturbance
Note: Not recommended for experiments involving phase contrast microscopy or immunofluorescence.
Thermanox™ coverslips provide excellent microscopic analysis with minimal cell disturbance
Note: Not recommended for experiments involving phase contrast microscopy or immunofluorescence.
Coverslips are 0.2mm thick with good resistance to most solvents used in electron microscopy. Coverslips may be cut easily without shattering. Radiation sterilized.
Ordering information: Packaged 50 per sleeve.
Customers Who Bought This Also Bought