Specifications
- Capacity:Absorbs up to 64.3 L (17 gal.)
- Environmentally Preferable:
- Cat. no.:56620-448
- Supplier No.:KIT228
Specifications
About this item
Safety-yellow, heavy-gauge, welded steel cabinet is printed "Spill Kit" for quick identification
Prepacked cabinet provides easy access to contents. See-through front panel lets you easily check inventory levels. Includes write-on label for emergency contact info. Two adjustable shelves.
PIG BLUE Socks stop spreading spills; PIG Mat and Pillows absorb quickly; wipers clean surfaces and hands.