PYREX® Hollow Full Length [ST] Stoppers, Corning
Supplier: Corning
Certificates
About this item
Hollow, full length stopper closed at the bottom from PYREX®, borosilicate glass.
- Interchangeable with standard taper joints
They are lightweight, but strong enough to keep chipping and breakage to a minimum. Barrel-shaped head is easy to clean and rotate, side grooves prevent slippage, and flat top allows stopper to stand on head.
Specifications
- Catalog No:
- 89091-610
- 59435-092
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey