Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Glycerine
Glycerol, CellPro™
Glycerol, CellPro™
104028-854
:
Alkali Scientific
:
Glycerol, CellPro™
104028-854
:
Alkali Scientific
:
DB0268
:
Pack type:
Plastic bottle
Size:
500 ml
Cat. no.:
104028-854
:
:
92.09 g/mol
:
1219
:
56-81-5
:
HOCH₂CH(OH)CH₂OH
:
MFCD00004722