Order Entry
Canada
ContactUsLinkComponent
CellPro™ Dextran Sulfate, 50%, Hybridization Grade, Alkali Scientific
CellPro™ Dextran Sulfate, 50%, Hybridization Grade, Alkali Scientific
Catalog # 104026-604
Supplier:  Alkali Scientific
CAS Number:  
undefined
CellPro™ Dextran Sulfate, 50%, Hybridization Grade, Alkali Scientific
Catalog # 104026-604
Supplier:  Alkali Scientific
Supplier Number:  DB0265
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • CAS No.:
    9011-18-1
  • Description:
    CellPro™ Dextran sulfate, 50%, Hybridization Grade
  • Size:
    100 ml
  • Cat. no.:
    104026-604

Specifications

About this item

Experience the excellence of CellPro™ Dextran Sulfate, meticulously crafted by Alkali Scientific™ to meet the stringent demands of hybridization assays.

  • Hybridization Grade
  • Sterile
  • Filtered

This high-quality Hybridization Grade Dextran Sulfate boasts a concentration of 50% and is supplied in a convenient 100mL volume. With its superior purity and performance, CellPro™ Dextran Sulfate ensures optimal results in various molecular biology applications, including nucleic acid hybridization and DNA-protein interactions. Trust CellPro™ Dextran Sulfate to deliver exceptional reliability and reproducibility, enabling precise and accurate experimental outcomes. Elevate your research with CellPro™ Dextran Sulfate and unlock new possibilities in molecular biology studies.

Caution: UN No. 3248