Sample Vials, with HDPE Caps
About this item
Polypropylene vials, with attached or separated HDPE Caps.
- Self-standing: the vial placed on the lab workbench steadily even without a base
- Sterilized by E-beam, Sterility Assurance Level: SAL 10⁻⁶
- Safe, fast, environment-friendly, no chemical residues
- DNAse-, RNAse-, pyrogen-,endotoxin-free
- Disposable
Excellent sealing performance, no leakage of negative pressure 95 Kpa, ensuring safe and effective air transportation.
Temperature range: Approx.–196 to 121 °C
Certifications: Meets USP VI standard.
13 Options Available Below
Product Details & Documents
Polypropylene vials, with attached or separated HDPE Caps.
- Self-standing: the vial placed on the lab workbench steadily even without a base
- Sterilized by E-beam, Sterility Assurance Level: SAL 10⁻⁶
- Safe, fast, environment-friendly, no chemical residues
- DNAse-, RNAse-, pyrogen-,endotoxin-free
- Disposable
Excellent sealing performance, no leakage of negative pressure 95 Kpa, ensuring safe and effective air transportation.
Temperature range: Approx.–196 to 121 °C
Certifications: Meets USP VI standard.
Product Family Options
With attached caps
Product Information
- CapacityCap includedCap typeSterilityPackagingAvailabilityPrice
- 77508-440Sample vialSample vial3 mlYesNormal capSterile50/pack; 1000/caseCase of 1,000$692.45Each$692.45Supplier Number: 624201
- 77508-302Sample vialSample vial4 mlYesNormal capSterile50/pack; 1000/caseCase of 1,000$692.45Each$692.45Supplier Number: 608201
- 77508-404Sample vialSample vial5 mlYesNormal capSterile50/pack; 1000/caseCase of 1,000$692.45Each$692.45Supplier Number: 619011
- 77508-408Sample vialSample vial5 mlYesDouble-threaded screw capSterile50/pack; 1000/caseCase of 1,000$692.45Each$692.45Supplier Number: 619101
- 77508-412Sample vialSample vial5 mlYesPink capture capSterile50/pack; 1000/caseCase of 1,000$692.45Each$692.45Supplier Number: 619201
- 77508-444Sample vialSample vial10 mlYesNormal capSterile50/pack; 1000/caseCase of 1,000$800.04Each$800.04Supplier Number: 625001
- 77508-448Shorter sample vialShorter sample vial10 mlYesNormal capSterile50/pack; 1000/caseCase of 1,000$800.04Each$800.04Supplier Number: 625011
With separated caps
Product Information
- CapacityCap includedCap typeSterilityPackagingAvailabilityPrice
- 77508-442Sample vialSample vial3 mlYesNormal capSterile200 tubes/bag, 200 caps/bagCase of 1,000$658.86Each$658.86Supplier Number: 624207
- 77508-304Sample vialSample vial4 mlYesNormal capSterile200 tubes/bag, 200 caps/bagCase of 1,000$658.86Each$658.86Supplier Number: 608207
- 77508-406Sample vialSample vial5 mlYesConcave/convex capSterile200 tubes/bag, 200 caps/bagCase of 1,000$658.86Each$658.86Supplier Number: 619017
- 77508-410Sample vialSample vial5 mlYesDouble-threaded screw capSterile200 tubes/bag, 200 caps/bagCase of 1,000$658.86Each$658.86Supplier Number: 619107
- 77508-446Sample vialSample vial10 mlYesNormal capSterile200 tubes/bag, 200 caps/bagCase of 1,000$759.67Each$759.67Supplier Number: 625007
- 77508-450Shorter sample vialShorter sample vial10 mlYesNormal capSterile200 tubes/bag, 200 caps/bagCase of 1,000$759.67Each$759.67Supplier Number: 625017