Microcentrifuge Tubes, Polypropylene
Supplier: NEST SCIENTIFIC
Certificates
About this item
Microcentrifuge tubes are commonly used to collect cells and separate biomacromolecules by centrifugation in biochemistry and other biological fields.
- Non-pyrogenic
- DNase/RNase free and deemed animal free
- Resistant to chemical corrosion and low temperature
- Clear graduations and frosted marking area
- Withstand temperature ranging from –80 to 121 ℃
Available in two colors: Amber and clear. Amber centrifuge tubes can also be used to store photosensitive samples and special chemiluminescence reagents.
Ultra-high centrifugal stability, centrifugal tolerance up to 30000×g. Special safelock cap design provides secure lid sealing. Tube cap can be opened or closed with one hand, easy to operate.
Sterilized by E-beam, Sterility Assurance Level: SAL 10⁻⁶. The product has been irradiated and dosimetrically released based on ANSI/AAMI/ISO 11137.
Certifications: Meets USP, Class VI standards.
Micro tube box
Product Information
Frequently Bought Together