Order Entry
Canada
Orders LinkContactUsLinkComponent
Disposable Sampler, ITM

Disposable Sampler, ITM

Disposable sampler, 2 ml ITM, 5 ml
Catalog # 77507-432
Supplier:   NEST SCIENTIFIC
Supplier #:  202135
Disposable Sampler, ITM
Catalog # 77507-432
Supplier:  NEST SCIENTIFIC
Disposable Sampler, ITM

Disposable sampler, 2 ml ITM, 5 ml

Price
$224.10
Log in for availability
Case of 100 Each
Product Details & Documents

5 ml vial with 2 ml ITM (Inactivation Transport Media).

Specifications
  • Volume:
    5 ml
  • Description:
    Disposable sampler, 2 ml ITM
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.