High Efficiency Erlenmeyer Flasks, Polycarbonate, Baffled Bottom
About this item
High-efficiency, large-volume culture flasks allow cells to show strong viability with large expression amount of proteins in the culture of mammalian cells and insect cells. During the culturing process, the use rate of the shaker is significantly increased, and the survival rate and viability of cells are both dramatically elevated. NEST culture flasks also provide high repeatability, which allows highly inter-batch consistency of cell growth and yield.
- PC bottle, which meets the USP Class 6 Standard, with high transparency, has strong impact resistance, oxidation resistance and can withstand a high temperature up to 121 ℃
- There’s a scale made by injection molding on the flask body to facilitate observing the filled liquid volume
- Non-Pyrogenic, DNase/Rnase free
- Electron beam sterilization, SAL= 10⁻⁶
Baffle-designed Erlenmeyer flasks promote optimal and consistent solution agitation when used with flask shakers.
Advantage of 3 L wide-mouth:
The bottom diameter of the 3 L wide-mouth high efficiency erlenmeyer flask is consistent with that of the 3 L high efficiency erlenmeyer flask, and its mouth diameter adopts the neck size of the 5 L high efficiency erlenmeyer flask, which not only saves space usage, but also increases oxygen flux, also increasing the efficiency of cell culture.
: Individually packaged.
8 Options Available Below
Product Details & Documents
High-efficiency, large-volume culture flasks allow cells to show strong viability with large expression amount of proteins in the culture of mammalian cells and insect cells. During the culturing process, the use rate of the shaker is significantly increased, and the survival rate and viability of cells are both dramatically elevated. NEST culture flasks also provide high repeatability, which allows highly inter-batch consistency of cell growth and yield.
- PC bottle, which meets the USP Class 6 Standard, with high transparency, has strong impact resistance, oxidation resistance and can withstand a high temperature up to 121 ℃
- There’s a scale made by injection molding on the flask body to facilitate observing the filled liquid volume
- Non-Pyrogenic, DNase/Rnase free
- Electron beam sterilization, SAL= 10⁻⁶
Baffle-designed Erlenmeyer flasks promote optimal and consistent solution agitation when used with flask shakers.
Advantage of 3 L wide-mouth:
The bottom diameter of the 3 L wide-mouth high efficiency erlenmeyer flask is consistent with that of the 3 L high efficiency erlenmeyer flask, and its mouth diameter adopts the neck size of the 5 L high efficiency erlenmeyer flask, which not only saves space usage, but also increases oxygen flux, also increasing the efficiency of cell culture.
: Individually packaged.
Product Family Options
With sealed cap
Product Information
- CapacitySterilityAvailabilityPrice
- 77508-870PC High efficient Erlenmeyer flask, baffledPC High efficient Erlenmeyer flask, baffled2 LSterileCase of 4$339.55Each$339.55Supplier Number: 785105
- 77508-886PC High efficient Erlenmeyer flask, baffledPC High efficient Erlenmeyer flask, baffled3 LSterileCase of 4$540.06Each$540.06Supplier Number: 786105
- 77508-894PC High efficient Erlenmeyer flask, baffled, wide mouthPC High efficient Erlenmeyer flask, baffled, wide mouth3 LSterileCase of 4$540.06Each$540.06Supplier Number: 786505
- 77508-906PC High efficient Erlenmeyer flask, baffledPC High efficient Erlenmeyer flask, baffled5 LSterileCase of 4$773.22Each$773.22Supplier Number: 787005
With vented cap
Product Information
- CapacitySterilityAvailabilityPrice
- 77508-874PC High efficient Erlenmeyer flask, baffledPC High efficient Erlenmeyer flask, baffled2 LSterileCase of 4$348.33Each$348.33Supplier Number: 785115
- 77508-890PC High efficient Erlenmeyer flask, baffledPC High efficient Erlenmeyer flask, baffled3 LSterileCase of 4$546.38Each$546.38Supplier Number: 786115
- 77508-898PC High efficient Erlenmeyer flask, baffled, wide mouthPC High efficient Erlenmeyer flask, baffled, wide mouth3 LSterileCase of 4$541.48Each$541.48Supplier Number: 786515
- 77508-910PC High efficient Erlenmeyer flask, baffledPC High efficient Erlenmeyer flask, baffled5 LSterileCase of 4$797.96Each$797.96Supplier Number: 787015