Orders LinkContactUsLinkComponent
Erlenmeyer Flasks, Polycarbonate
Supplier:   NEST SCIENTIFIC
Erlenmeyer Flasks, Polycarbonate
Supplier:  NEST SCIENTIFIC
Certificates
Erlenmeyer Flasks, Polycarbonate
16 Options Available Below
Product Details & Documents
PC Erlenmeyer flask with flat or baffled bottom are equipped with seal caps or vent filter caps. The whole collection of Erlenmeyer flasks are sterile and enzyme-free, and are intended for the culture of suspended cells and bacteria.
  • PC bottle, which meets the USP Class 6 standard, with high transparency, has strong impact resistance, oxidation resistance and can withstand a high temperature up to 121 ℃
  • There is a scale made by injection molding on the flask body to facilitate observing the filled liquid volume
  • Non-pyrogenic, DNase/Rnase free
  • Electron beam sterilization, SAL= 10⁻⁶
  • Individually packaged
In the culture process of mammalian cells and insect cells, the cells show strong viability and high protein expression in Erlenmeyer flasks, baffled. The groove at the bottom forms a vortex that accelerates the reagent flow to improve the oxygen exchange rate between the surface and the outside for higher dissolved oxygen content.
Baffle-designed erlenmeyer flasks promote optimal and consistent solution agitation when used with flask shakers.
 : ISO 9001 and 13485 certified.
Product Family Options
Baffled bottom
Product Information
  • Capacity
    Sterility
    Height
    Neck O.D.
    Cap type
    Availability
    Price
  • 77508-812
    Conical Erlenmeyer flasks, Polycarbonate, 1 per pack, 24 per case
    Conical Erlenmeyer flasks, Polycarbonate, 1 per pack, 24 per case
    125 ml
    Sterile
    Plug seal cap
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $238.59
    $238.59
    Log in for availability
    Each
    Each - $238.59
    $238.59
    Case of 24
       $238.59
    Each
       $238.59
    Supplier Number: 781201
  • 77508-814
    Conical Erlenmeyer flasks, Polycarbonate, 1 per pack, 24 per case
    Conical Erlenmeyer flasks, Polycarbonate, 1 per pack, 24 per case
    125 ml
    Sterile
    Vent cap
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $248.76
    $248.76
    Log in for availability
    Each
    Each - $248.76
    $248.76
    Case of 24
       $248.76
    Each
       $248.76
    Supplier Number: 781211
  • 77508-828
    Conical Erlenmeyer flasks, Polycarbonate, 1 per pack, 12 per case
    Conical Erlenmeyer flasks, Polycarbonate, 1 per pack, 12 per case
    250 ml
    Sterile
    Plug seal cap
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $164.22
    $164.22
    Log in for availability
    Each
    Each - $164.22
    $164.22
    Case of 12
       $164.22
    Each
       $164.22
    Supplier Number: 782201
  • 77508-830
    Conical Erlenmeyer flasks, Polycarbonate, 1 per pack, 12 per case
    Conical Erlenmeyer flasks, Polycarbonate, 1 per pack, 12 per case
    250 ml
    Sterile
    Vent cap
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $174.09
    $174.09
    Log in for availability
    Each
    Each - $174.09
    $174.09
    Case of 12
       $174.09
    Each
       $174.09
    Supplier Number: 782211
  • 77508-844
    Conical Erlenmeyer flasks, Polycarbonate, 1 per pack, 12 per case
    Conical Erlenmeyer flasks, Polycarbonate, 1 per pack, 12 per case
    500 ml
    Sterile
    Plug seal cap
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $231.69
    $231.69
    Log in for availability
    Each
    Each - $231.69
    $231.69
    Case of 12
       $231.69
    Each
       $231.69
    Supplier Number: 783201
  • 77508-846
    Conical Erlenmeyer flasks, Polycarbonate, 1 per pack, 12 per case
    Conical Erlenmeyer flasks, Polycarbonate, 1 per pack, 12 per case
    500 ml
    Sterile
    Vent cap
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $253.22
    $253.22
    Log in for availability
    Each
    Each - $253.22
    $253.22
    Case of 12
       $253.22
    Each
       $253.22
    Supplier Number: 783211
  • 77508-860
    Conical Erlenmeyer flasks, Polycarbonate, 1 per pack, 6 per case
    Conical Erlenmeyer flasks, Polycarbonate, 1 per pack, 6 per case
    1000 ml
    Sterile
    Plug seal cap
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 6
    Case of 6 - $176.03
    $176.03
    Log in for availability
    Each
    Each - $176.03
    $176.03
    Case of 6
       $176.03
    Each
       $176.03
    Supplier Number: 784201
  • 77508-862
    Conical Erlenmeyer flasks, Polycarbonate, 1 per pack, 6 per case
    Conical Erlenmeyer flasks, Polycarbonate, 1 per pack, 6 per case
    1000 ml
    Sterile
    Vent cap
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 6
    Case of 6 - $185.85
    $185.85
    Log in for availability
    Each
    Each - $185.85
    $185.85
    Case of 6
       $185.85
    Each
       $185.85
    Supplier Number: 784211
Flat bottom
Product Information
  • Capacity
    Sterility
    Height
    Neck O.D.
    Cap type
    Availability
    Price
  • 77508-808
    Conical Erlenmeyer flasks, Polycarbonate, Bottom Ø: 66 mm, 1 per pack, 24 per case
    Conical Erlenmeyer flasks, Polycarbonate, Bottom Ø: 66 mm, 1 per pack, 24 per case
    125 ml
    Sterile
    106.8 mm
    34 mm
    Plug seal cap
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $218.20
    $218.20
    Log in for availability
    Each
    Each - $218.20
    $218.20
    Case of 24
       $218.20
    Each
       $218.20
    Supplier Number: 781101
  • 77508-810
    Conical Erlenmeyer flasks, Polycarbonate, Bottom Ø: 66 mm, 1 per pack, 24 per case
    Conical Erlenmeyer flasks, Polycarbonate, Bottom Ø: 66 mm, 1 per pack, 24 per case
    125 ml
    Sterile
    106.8 mm
    34 mm
    Vent cap
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $220.20
    $220.20
    Log in for availability
    Each
    Each - $220.20
    $220.20
    Case of 24
       $220.20
    Each
       $220.20
    Supplier Number: 781111
  • 77508-824
    Conical Erlenmeyer flasks, Polycarbonate, Bottom Ø: 83 mm, 1 per pack, 12 per case
    Conical Erlenmeyer flasks, Polycarbonate, Bottom Ø: 83 mm, 1 per pack, 12 per case
    250 ml
    Sterile
    137.8 mm
    34 mm
    Plug seal cap
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $149.40
    $149.40
    Log in for availability
    Each
    Each - $149.40
    $149.40
    Case of 12
       $149.40
    Each
       $149.40
    Supplier Number: 782101
  • 77508-826
    Conical Erlenmeyer flasks, Polycarbonate, Bottom Ø: 83 mm, 1 per pack, 12 per case
    Conical Erlenmeyer flasks, Polycarbonate, Bottom Ø: 83 mm, 1 per pack, 12 per case
    250 ml
    Sterile
    137.8 mm
    34 mm
    Vent cap
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $170.32
    $170.32
    Log in for availability
    Each
    Each - $170.32
    $170.32
    Case of 12
       $170.32
    Each
       $170.32
    Supplier Number: 782111
  • 77508-840
    Conical Erlenmeyer flasks, Polycarbonate, Bottom Ø: 101 mm, 1 per pack, 12 per case
    Conical Erlenmeyer flasks, Polycarbonate, Bottom Ø: 101 mm, 1 per pack, 12 per case
    500 ml
    Sterile
    175.8 mm
    39 mm
    Plug seal cap
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $219.14
    $219.14
    Log in for availability
    Each
    Each - $219.14
    $219.14
    Case of 12
       $219.14
    Each
       $219.14
    Supplier Number: 783101
  • 77508-842
    Conical Erlenmeyer flasks, Polycarbonate, Bottom Ø: 101 mm, 1 per pack, 12 per case
    Conical Erlenmeyer flasks, Polycarbonate, Bottom Ø: 101 mm, 1 per pack, 12 per case
    500 ml
    Sterile
    175.8 mm
    39 mm
    Vent cap
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $231.69
    $231.69
    Log in for availability
    Each
    Each - $231.69
    $231.69
    Case of 12
       $231.69
    Each
       $231.69
    Supplier Number: 783111
  • 77508-856
    Conical Erlenmeyer flasks, Polycarbonate, Bottom Ø: 127 mm, 1 per pack, 6 per case
    Conical Erlenmeyer flasks, Polycarbonate, Bottom Ø: 127 mm, 1 per pack, 6 per case
    1000 ml
    Sterile
    213.3 mm
    39 mm
    Plug seal cap
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 6
    Case of 6 - $166.20
    $166.20
    Log in for availability
    Each
    Each - $166.20
    $166.20
    Case of 6
       $166.20
    Each
       $166.20
    Supplier Number: 784101
  • 77508-858
    Conical Erlenmeyer flasks, Polycarbonate, Bottom Ø: 127 mm, 1 per pack, 6 per case
    Conical Erlenmeyer flasks, Polycarbonate, Bottom Ø: 127 mm, 1 per pack, 6 per case
    1000 ml
    Sterile
    213.3 mm
    39 mm
    Vent cap
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 6
    Case of 6 - $178.30
    $178.30
    Log in for availability
    Each
    Each - $178.30
    $178.30
    Case of 6
       $178.30
    Each
       $178.30
    Supplier Number: 784111